Assay Details
| Gene Symbol: SIRPG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB042624.1 | - | 1 - 2 | 156 | 83 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AB042624.1 transcripts.
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000216927.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000216927.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000216927.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000216927.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000303415.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000303415.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000303415.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000303415.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000303415.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000303415.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000303415.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000303415.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000344103.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000344103.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000344103.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000344103.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000344103.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000344103.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000381580.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000381580.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000381580.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000381580.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000381580.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000381580.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000381580.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000381583.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000381583.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000381583.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000381583.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000381583.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000381583.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000381583.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000478145.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000478145.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000478145.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000478145.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000497407.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000497407.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | ENST00000497407.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | NM_001039508.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | NM_001039508.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | NM_001039508.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | NM_001039508.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | NM_001039508.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | NM_001039508.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | NM_001039508.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | NM_018556.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | NM_018556.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | NM_018556.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | NM_018556.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | NM_018556.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | NM_018556.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | NM_018556.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | NM_018556.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | NM_080816.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | NM_080816.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | NM_080816.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | NM_080816.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | NM_080816.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000009971.hg.1 | NM_080816.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000216927.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000216927.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000216927.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000216927.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000303415.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000303415.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000303415.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000303415.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000303415.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000303415.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000303415.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000303415.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000344103.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000344103.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000344103.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000344103.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000344103.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000344103.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000381580.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000381580.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000381580.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000381580.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000381580.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000381580.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000381580.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000381583.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000381583.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000381583.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000381583.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000381583.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000381583.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000381583.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000478145.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000478145.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000478145.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000478145.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000497407.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000497407.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | ENST00000497407.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | NM_001039508.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | NM_001039508.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | NM_001039508.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | NM_001039508.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | NM_001039508.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | NM_001039508.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | NM_001039508.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | NM_018556.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | NM_018556.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | NM_018556.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | NM_018556.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | NM_018556.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | NM_018556.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | NM_018556.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | NM_018556.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | NM_080816.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | NM_080816.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | NM_080816.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | NM_080816.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | NM_080816.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000009971.hg.1 | NM_080816.2 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 206705 | - | - |

SIRPG
Genomic Map