Assay Details
| Gene Symbol: OSGIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY258067.1 | - | 1 - 2 | 440 | 151 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AY258067.1 transcripts.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000561552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000561552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000561552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000561552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000561552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000561552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000561552.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000563543.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000563543.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000563543.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000565123.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000565123.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000565123.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000565123.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000565123.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000565123.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000565123.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000565123.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000567707.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000567707.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000567707.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | ENST00000567707.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011440.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000343939.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000361711.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000393306.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000561552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000561552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000561552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000561552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000561552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000561552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000561552.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000563543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000563543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000563543.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000565123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000565123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000565123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000565123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000565123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000565123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000565123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000565123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000567707.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000567707.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000567707.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | ENST00000567707.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | NM_182981.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011440.hg.1 | NM_182981.2 | Refseq |

OSGIN1
Genomic Map