Assay Details
| Gene Symbol: FGFR1OP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007045.3 | NP_008976.1 | 6 - 7 | 610 | 91 | |
| GenBank mRNA | AK312791.1 | - | 6 - 7 | 521 | 91 | |
| Y18046.1 | - | 6 - 7 | 599 | 91 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06004105.hg.1
Set Membership: |
|

FGFR1OP
Genomic Map