Assay Details
| Gene Symbol: DYRK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003583.3 | NP_003574.1 | 1 - 2 | 451 | 89 | |
| GenBank mRNA | BC005809.2 | - | 1 - 2 | 405 | 3831977 | 89 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12000597.hg
Additional Information:
This assay is in the 5'UTR region of the BC005809.2,NM_003583.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
cellular response to DNA damage stimulus
smoothened signaling pathway
peptidyl-tyrosine phosphorylation
intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
positive regulation of glycogen biosynthetic process
negative regulation of NFAT protein import into nucleus
regulation of signal transduction by p53 class mediator
magnesium ion binding
protein serine/threonine kinase activity
protein serine/threonine/tyrosine kinase activity
protein tyrosine kinase activity
protein binding
ATP binding
manganese ion binding
ubiquitin binding
protein phosphorylation
cellular response to DNA damage stimulus
smoothened signaling pathway
peptidyl-tyrosine phosphorylation
intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
positive regulation of glycogen biosynthetic process
negative regulation of NFAT protein import into nucleus
regulation of signal transduction by p53 class mediator
magnesium ion binding
protein serine/threonine kinase activity
protein serine/threonine/tyrosine kinase activity
protein tyrosine kinase activity
protein binding
ATP binding
manganese ion binding
ubiquitin binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR12007868.hg.1 | ENST00000344096.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12007868.hg.1 | NM_003583.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12007868.hg.1 | NM_006482.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12007868.hg.1 | TC12000597.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12007872.hg.1 | ENST00000344096.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12007872.hg.1 | ENST00000393555.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12007872.hg.1 | NM_003583.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12007872.hg.1 | NM_006482.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12007872.hg.1 | TC12000597.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008081.hg.1 | ENST00000319833.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008081.hg.1 | ENST00000319833.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008081.hg.1 | ENST00000319833.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008081.hg.1 | ENST00000344096.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008081.hg.1 | ENST00000344096.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008081.hg.1 | ENST00000344096.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008081.hg.1 | ENST00000344096.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008081.hg.1 | ENST00000344096.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008081.hg.1 | ENST00000344096.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008081.hg.1 | ENST00000344096.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008081.hg.1 | ENST00000344096.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008081.hg.1 | ENST00000344096.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008081.hg.1 | ENST00000344096.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008081.hg.1 | ENST00000344096.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008081.hg.1 | ENST00000344096.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008081.hg.1 | ENST00000344096.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008081.hg.1 | ENST00000344096.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008081.hg.1 | ENST00000393555.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008081.hg.1 | ENST00000393555.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008081.hg.1 | ENST00000393555.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008081.hg.1 | ENST00000393555.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008081.hg.1 | ENST00000393555.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008081.hg.1 | ENST00000393555.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008081.hg.1 | ENST00000393555.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008081.hg.1 | ENST00000393555.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008081.hg.1 | ENST00000393555.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008081.hg.1 | ENST00000393555.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008081.hg.1 | ENST00000537632.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008081.hg.1 | ENST00000537632.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008081.hg.1 | ENST00000537632.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008081.hg.1 | ENST00000542503.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008081.hg.1 | ENST00000542503.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008081.hg.1 | ENST00000542503.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008081.hg.1 | ENST00000543747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008081.hg.1 | ENST00000543747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008081.hg.1 | ENST00000543747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008081.hg.1 | ENST00000543747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008081.hg.1 | NM_003583.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008081.hg.1 | NM_003583.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008081.hg.1 | NM_003583.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008081.hg.1 | NM_003583.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008081.hg.1 | NM_003583.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008081.hg.1 | NM_003583.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008081.hg.1 | NM_003583.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008081.hg.1 | NM_003583.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008081.hg.1 | NM_003583.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008081.hg.1 | NM_003583.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008081.hg.1 | NM_003583.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008081.hg.1 | NM_003583.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008081.hg.1 | NM_006482.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008081.hg.1 | NM_006482.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008081.hg.1 | NM_006482.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008081.hg.1 | NM_006482.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008081.hg.1 | NM_006482.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008081.hg.1 | NM_006482.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008081.hg.1 | NM_006482.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008081.hg.1 | NM_006482.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008081.hg.1 | NM_006482.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008081.hg.1 | NM_006482.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008081.hg.1 | NM_006482.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008081.hg.1 | NM_006482.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008081.hg.1 | NM_006482.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12007873.hg.1 | ENST00000344096.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12007873.hg.1 | ENST00000393555.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12007873.hg.1 | ENST00000537632.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12007873.hg.1 | NM_003583.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12007873.hg.1 | NM_006482.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12007873.hg.1 | TC12000597.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12007870.hg.1 | ENST00000344096.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12007870.hg.1 | ENST00000393555.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12007870.hg.1 | NM_003583.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12007870.hg.1 | NM_006482.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12007870.hg.1 | TC12000597.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12007871.hg.1 | ENST00000344096.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12007871.hg.1 | ENST00000393555.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12007871.hg.1 | NM_003583.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12007871.hg.1 | NM_006482.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12007871.hg.1 | TC12000597.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12007869.hg.1 | ENST00000344096.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12007869.hg.1 | ENST00000393555.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12007869.hg.1 | NM_003583.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12007869.hg.1 | NM_006482.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12007869.hg.1 | TC12000597.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1200159125.hg.1 | ENST00000344096.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200159125.hg.1 | NM_003583.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200159125.hg.1 | NM_006482.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200159125.hg.1 | ENST00000344096.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200159125.hg.1 | NM_003583.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200159125.hg.1 | NM_006482.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200159125.hg.1 | TC1200008081.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200159129.hg.1 | ENST00000344096.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200159129.hg.1 | ENST00000393555.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200159129.hg.1 | NM_003583.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200159129.hg.1 | NM_006482.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200159129.hg.1 | ENST00000344096.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200159129.hg.1 | ENST00000393555.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200159129.hg.1 | NM_003583.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200159129.hg.1 | NM_006482.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200159129.hg.1 | TC1200008081.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200159131.hg.1 | ENST00000344096.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200159131.hg.1 | ENST00000393555.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200159131.hg.1 | ENST00000537632.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200159131.hg.1 | NM_003583.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200159131.hg.1 | NM_006482.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200159131.hg.1 | ENST00000344096.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200159131.hg.1 | ENST00000393555.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200159131.hg.1 | ENST00000537632.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200159131.hg.1 | NM_003583.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200159131.hg.1 | NM_006482.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200159131.hg.1 | TC1200008081.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200159126.hg.1 | ENST00000344096.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200159126.hg.1 | ENST00000393555.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200159126.hg.1 | NM_003583.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200159126.hg.1 | NM_006482.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200159126.hg.1 | ENST00000344096.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200159126.hg.1 | ENST00000393555.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200159126.hg.1 | NM_003583.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200159126.hg.1 | NM_006482.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200159126.hg.1 | TC1200008081.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200008081.hg.1 | ENST00000319833.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008081.hg.1 | ENST00000319833.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008081.hg.1 | ENST00000319833.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008081.hg.1 | ENST00000344096.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008081.hg.1 | ENST00000344096.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008081.hg.1 | ENST00000344096.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008081.hg.1 | ENST00000344096.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008081.hg.1 | ENST00000344096.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008081.hg.1 | ENST00000344096.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008081.hg.1 | ENST00000344096.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008081.hg.1 | ENST00000344096.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008081.hg.1 | ENST00000344096.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008081.hg.1 | ENST00000344096.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008081.hg.1 | ENST00000344096.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008081.hg.1 | ENST00000344096.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008081.hg.1 | ENST00000344096.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008081.hg.1 | ENST00000344096.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008081.hg.1 | ENST00000393555.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008081.hg.1 | ENST00000393555.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008081.hg.1 | ENST00000393555.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008081.hg.1 | ENST00000393555.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008081.hg.1 | ENST00000393555.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008081.hg.1 | ENST00000393555.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008081.hg.1 | ENST00000393555.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008081.hg.1 | ENST00000393555.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008081.hg.1 | ENST00000393555.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008081.hg.1 | ENST00000393555.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008081.hg.1 | ENST00000537632.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008081.hg.1 | ENST00000537632.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008081.hg.1 | ENST00000537632.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008081.hg.1 | ENST00000542503.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008081.hg.1 | ENST00000542503.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008081.hg.1 | ENST00000542503.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008081.hg.1 | ENST00000543747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008081.hg.1 | ENST00000543747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008081.hg.1 | ENST00000543747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008081.hg.1 | ENST00000543747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008081.hg.1 | NM_003583.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008081.hg.1 | NM_003583.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008081.hg.1 | NM_003583.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008081.hg.1 | NM_003583.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008081.hg.1 | NM_003583.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008081.hg.1 | NM_003583.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008081.hg.1 | NM_003583.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008081.hg.1 | NM_003583.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008081.hg.1 | NM_003583.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008081.hg.1 | NM_003583.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008081.hg.1 | NM_003583.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008081.hg.1 | NM_003583.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008081.hg.1 | NM_006482.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008081.hg.1 | NM_006482.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008081.hg.1 | NM_006482.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008081.hg.1 | NM_006482.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008081.hg.1 | NM_006482.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008081.hg.1 | NM_006482.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008081.hg.1 | NM_006482.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008081.hg.1 | NM_006482.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008081.hg.1 | NM_006482.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008081.hg.1 | NM_006482.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008081.hg.1 | NM_006482.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008081.hg.1 | NM_006482.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008081.hg.1 | NM_006482.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200159128.hg.1 | ENST00000344096.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200159128.hg.1 | ENST00000393555.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200159128.hg.1 | NM_003583.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200159128.hg.1 | NM_006482.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200159128.hg.1 | ENST00000344096.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200159128.hg.1 | ENST00000393555.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200159128.hg.1 | NM_003583.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200159128.hg.1 | NM_006482.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200159128.hg.1 | TC1200008081.hg.1 | Clariom D TC |

DYRK2
Genomic Map