Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ras homolog family member D | RHOD |
ARHD, RHOHP1, RHOM, Rho |
-121.0kb | Sense | Chr. 11 - 67056818 - 67072017 on NCBI Build GRCh38 |
| Downstream | G protein-coupled receptor kinase 2 | GRK2 |
ADRBK1, BARK1, BETA-ARK1 |
71.0kb | Sense | Chr. 11 - 67266420 - 67286558 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

Genomic Map