| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19014125.hg.1 |
ENST00000221566.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19014125.hg.1 |
NM_003021.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19014125.hg.1 |
TC19001036.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009254.hg.1 |
ENST00000221566.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009254.hg.1 |
ENST00000221566.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009254.hg.1 |
ENST00000221566.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009254.hg.1 |
ENST00000221566.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009254.hg.1 |
ENST00000221566.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009254.hg.1 |
ENST00000221566.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009254.hg.1 |
ENST00000221566.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009254.hg.1 |
ENST00000221566.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009254.hg.1 |
ENST00000221566.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009254.hg.1 |
ENST00000221566.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009254.hg.1 |
ENST00000221566.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009254.hg.1 |
ENST00000221566.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009254.hg.1 |
ENST00000221566.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009254.hg.1 |
ENST00000586711.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009254.hg.1 |
ENST00000587866.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009254.hg.1 |
ENST00000587866.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009254.hg.1 |
ENST00000587866.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009254.hg.1 |
ENST00000589251.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009254.hg.1 |
ENST00000589251.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009254.hg.1 |
ENST00000589251.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009254.hg.1 |
ENST00000589251.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009254.hg.1 |
ENST00000589251.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009254.hg.1 |
ENST00000589251.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009254.hg.1 |
ENST00000591984.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009254.hg.1 |
ENST00000591984.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009254.hg.1 |
ENST00000591984.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009254.hg.1 |
ENST00000591984.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009254.hg.1 |
ENST00000591984.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009254.hg.1 |
NM_003021.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009254.hg.1 |
NM_003021.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009254.hg.1 |
NM_003021.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009254.hg.1 |
NM_003021.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009254.hg.1 |
NM_003021.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009254.hg.1 |
NM_003021.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009254.hg.1 |
NM_003021.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009254.hg.1 |
NM_003021.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009254.hg.1 |
NM_003021.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009254.hg.1 |
NM_003021.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009254.hg.1 |
NM_003021.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009254.hg.1 |
NM_003021.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009254.hg.1 |
NM_003021.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900173738.hg.1 |
ENST00000221566.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900173738.hg.1 |
NM_003021.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900173738.hg.1 |
TC1900009254.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900173738.hg.1 |
ENST00000221566.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900173738.hg.1 |
NM_003021.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009254.hg.1 |
ENST00000221566.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009254.hg.1 |
ENST00000221566.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009254.hg.1 |
ENST00000221566.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009254.hg.1 |
ENST00000221566.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009254.hg.1 |
ENST00000221566.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009254.hg.1 |
ENST00000221566.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009254.hg.1 |
ENST00000221566.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009254.hg.1 |
ENST00000221566.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009254.hg.1 |
ENST00000221566.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009254.hg.1 |
ENST00000221566.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009254.hg.1 |
ENST00000221566.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009254.hg.1 |
ENST00000221566.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009254.hg.1 |
ENST00000221566.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009254.hg.1 |
ENST00000586711.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009254.hg.1 |
ENST00000587866.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009254.hg.1 |
ENST00000587866.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009254.hg.1 |
ENST00000587866.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009254.hg.1 |
ENST00000589251.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009254.hg.1 |
ENST00000589251.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009254.hg.1 |
ENST00000589251.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009254.hg.1 |
ENST00000589251.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009254.hg.1 |
ENST00000589251.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009254.hg.1 |
ENST00000589251.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009254.hg.1 |
ENST00000591984.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009254.hg.1 |
ENST00000591984.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009254.hg.1 |
ENST00000591984.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009254.hg.1 |
ENST00000591984.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009254.hg.1 |
ENST00000591984.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009254.hg.1 |
NM_003021.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009254.hg.1 |
NM_003021.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009254.hg.1 |
NM_003021.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009254.hg.1 |
NM_003021.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009254.hg.1 |
NM_003021.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009254.hg.1 |
NM_003021.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009254.hg.1 |
NM_003021.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009254.hg.1 |
NM_003021.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009254.hg.1 |
NM_003021.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009254.hg.1 |
NM_003021.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009254.hg.1 |
NM_003021.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009254.hg.1 |
NM_003021.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009254.hg.1 |
NM_003021.3 |
Refseq |