Assay Details
| Gene Symbol: RUBCN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145642.3 | NP_001139114.1 | 12 - 13 | 2197 | 64 | |
| NM_014687.2 | NP_055502.1 | 11 - 12 | 1993 | 64 | ||
| XM_005269374.3 | XP_005269431.1 | 12 - 13 | 2033 | 64 | ||
| XM_017007544.1 | XP_016863033.1 | 11 - 12 | 1879 | 64 | ||
| XM_017007545.1 | XP_016863034.1 | 11 - 12 | 2195 | 64 | ||
| GenBank mRNA | D86979.3 | - | 12 - 13 | 1886 | 64 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03002182.hg.1
Set Membership: |
|

RUBCN
Genomic Map