Assay Details
| Gene Symbol: ACBD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006717528.3 | XP_006717591.3 | 6 - 7 | 7111 | 131 | |
| XM_006717530.3 | XP_006717593.1 | 6 - 7 | 710 | 131 | ||
| XM_006717531.2 | XP_006717594.1 | 6 - 7 | 720 | 131 | ||
| XM_006717535.2 | XP_006717598.1 | 6 - 7 | 577 | 131 | ||
| XM_017016884.1 | XP_016872373.1 | 6 - 7 | 7111 | 131 | ||
| XM_017016887.1 | XP_016872376.1 | 6 - 7 | 641 | 131 | ||
| XM_017016888.1 | XP_016872377.1 | 6 - 7 | 706 | 131 | ||
| XM_017016890.1 | XP_016872379.1 | 6 - 7 | 7111 | 131 | ||
| XM_017016895.1 | XP_016872384.1 | 6 - 7 | 577 | 131 | ||
| XM_017016897.1 | XP_016872386.1 | 6 - 7 | 706 | 131 | ||
| XM_017016900.1 | XP_016872389.1 | 5 - 6 | 495 | 131 | ||
| XM_017016901.1 | XP_016872390.1 | 5 - 6 | 475 | 131 | ||
| XM_017016902.1 | XP_016872391.1 | 5 - 6 | 494 | 131 | ||
| XM_017016903.1 | XP_016872392.1 | 5 - 6 | 479 | 131 | ||
More Information
Set Membership: |
|

ACBD5
Genomic Map