Assay Details
| Gene Symbol: MANSC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018050.2 | NP_060520.2 | 1 - 2 | 158 | 81 | |
| XM_017019531.1 | XP_016875020.1 | 1 - 2 | 464 | 81 | ||
| GenBank mRNA | AK023622.1 | - | 1 - 2 | 158 | 81 | |
| AY358563.1 | - | 1 - 2 | 125 | 81 | ||
| BC032998.2 | - | 1 - 2 | 175 | 4824116 | 81 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12001253.hg
Additional Information:
This assay is in the 5'UTR region of the AK023622.1,AY358563.1,BC032998.2,NM_018050.2,XM_017019531.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR12016386.hg.1 | ENST00000535902.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12016386.hg.1 | NM_018050.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12016386.hg.1 | TC12001253.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12016388.hg.1 | ENST00000355566.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12016388.hg.1 | ENST00000396349.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12016388.hg.1 | ENST00000535902.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12016388.hg.1 | NM_018050.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12016388.hg.1 | TC12001253.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009964.hg.1 | ENST00000396349.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009964.hg.1 | ENST00000396349.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009964.hg.1 | ENST00000396349.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009964.hg.1 | ENST00000396349.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009964.hg.1 | ENST00000396349.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009964.hg.1 | ENST00000396349.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009964.hg.1 | ENST00000396349.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009964.hg.1 | ENST00000396349.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009964.hg.1 | ENST00000396349.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009964.hg.1 | ENST00000535902.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009964.hg.1 | ENST00000535902.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009964.hg.1 | ENST00000535902.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009964.hg.1 | ENST00000535902.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009964.hg.1 | ENST00000535902.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009964.hg.1 | ENST00000535902.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009964.hg.1 | ENST00000535902.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009964.hg.1 | ENST00000535902.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009964.hg.1 | ENST00000535902.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009964.hg.1 | ENST00000535902.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009964.hg.1 | ENST00000535902.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009964.hg.1 | ENST00000535902.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009964.hg.1 | ENST00000543314.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009964.hg.1 | ENST00000543314.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009964.hg.1 | ENST00000543314.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009964.hg.1 | ENST00000543314.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009964.hg.1 | ENST00000543314.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009964.hg.1 | ENST00000543314.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009964.hg.1 | ENST00000545735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009964.hg.1 | ENST00000545735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009964.hg.1 | ENST00000545735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009964.hg.1 | ENST00000545735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009964.hg.1 | NM_018050.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009964.hg.1 | NM_018050.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009964.hg.1 | NM_018050.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009964.hg.1 | NM_018050.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009964.hg.1 | NM_018050.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009964.hg.1 | NM_018050.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009964.hg.1 | NM_018050.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009964.hg.1 | NM_018050.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009964.hg.1 | NM_018050.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009964.hg.1 | NM_018050.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12016387.hg.1 | ENST00000396349.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12016387.hg.1 | ENST00000535902.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12016387.hg.1 | NM_018050.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12016387.hg.1 | TC12001253.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1200175272.hg.1 | ENST00000535902.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200175272.hg.1 | ENST00000543314.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200175272.hg.1 | NM_018050.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200175272.hg.1 | ENST00000535902.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200175272.hg.1 | ENST00000543314.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200175272.hg.1 | NM_018050.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200175272.hg.1 | TC1200009964.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200009964.hg.1 | ENST00000396349.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009964.hg.1 | ENST00000396349.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009964.hg.1 | ENST00000396349.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009964.hg.1 | ENST00000396349.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009964.hg.1 | ENST00000396349.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009964.hg.1 | ENST00000396349.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009964.hg.1 | ENST00000396349.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009964.hg.1 | ENST00000396349.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009964.hg.1 | ENST00000396349.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009964.hg.1 | ENST00000535902.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009964.hg.1 | ENST00000535902.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009964.hg.1 | ENST00000535902.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009964.hg.1 | ENST00000535902.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009964.hg.1 | ENST00000535902.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009964.hg.1 | ENST00000535902.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009964.hg.1 | ENST00000535902.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009964.hg.1 | ENST00000535902.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009964.hg.1 | ENST00000535902.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009964.hg.1 | ENST00000535902.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009964.hg.1 | ENST00000535902.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009964.hg.1 | ENST00000535902.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009964.hg.1 | ENST00000543314.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009964.hg.1 | ENST00000543314.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009964.hg.1 | ENST00000543314.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009964.hg.1 | ENST00000543314.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009964.hg.1 | ENST00000543314.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009964.hg.1 | ENST00000543314.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009964.hg.1 | ENST00000545735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009964.hg.1 | ENST00000545735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009964.hg.1 | ENST00000545735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009964.hg.1 | ENST00000545735.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009964.hg.1 | NM_018050.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009964.hg.1 | NM_018050.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009964.hg.1 | NM_018050.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009964.hg.1 | NM_018050.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009964.hg.1 | NM_018050.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009964.hg.1 | NM_018050.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009964.hg.1 | NM_018050.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009964.hg.1 | NM_018050.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009964.hg.1 | NM_018050.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009964.hg.1 | NM_018050.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200175274.hg.1 | ENST00000396349.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200175274.hg.1 | ENST00000535902.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200175274.hg.1 | ENST00000543314.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200175274.hg.1 | NM_018050.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200175274.hg.1 | ENST00000396349.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200175274.hg.1 | ENST00000535902.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200175274.hg.1 | ENST00000543314.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200175274.hg.1 | NM_018050.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200175274.hg.1 | TC1200009964.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200175273.hg.1 | ENST00000396349.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200175273.hg.1 | ENST00000535902.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200175273.hg.1 | ENST00000543314.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200175273.hg.1 | NM_018050.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200175273.hg.1 | ENST00000396349.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200175273.hg.1 | ENST00000535902.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200175273.hg.1 | ENST00000543314.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200175273.hg.1 | NM_018050.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200175273.hg.1 | TC1200009964.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200175276.hg.1 | ENST00000396349.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200175276.hg.1 | ENST00000535902.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200175276.hg.1 | ENST00000543314.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200175276.hg.1 | NM_018050.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200175276.hg.1 | ENST00000396349.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200175276.hg.1 | ENST00000535902.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200175276.hg.1 | ENST00000543314.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200175276.hg.1 | NM_018050.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200175276.hg.1 | TC1200009964.hg.1 | Clariom D TC |

MANSC1
Genomic Map