Assay Details
| Gene Symbol: SFMBT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016329.3 | NP_057413.2 | 10 - 11 | 1518 | 67 | |
| XM_005265221.3 | XP_005265278.1 | 10 - 11 | 1634 | 67 | ||
| XM_006713203.3 | XP_006713266.1 | 11 - 12 | 1756 | 67 | ||
| XM_006713204.3 | XP_006713267.1 | 10 - 11 | 1456 | 67 | ||
| XM_011533824.2 | XP_011532126.1 | 11 - 12 | 1632 | 67 | ||
| XM_011533825.2 | XP_011532127.1 | 11 - 12 | 1479 | 67 | ||
| GenBank mRNA | AB189472.1 | - | 9 - 10 | 1142 | 67 | |
| AF168132.1 | - | 10 - 11 | 1455 | 67 | ||
| AK313965.1 | - | 10 - 11 | 1513 | 67 | ||
| AL080140.1 | - | 7 - 8 | 796 | 67 | ||
| AL523328.3 | - | 5 - 6 | 545 | 67 | ||
| BC014614.2 | - | 10 - 11 | 1402 | 3029598 | 67 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03001464.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000470575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000470575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000470575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000470575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000470575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000470575.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000474837.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000474837.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000474837.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000474837.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000474837.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000483069.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000483069.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000483069.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000483069.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000483069.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000483069.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000483069.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000485494.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000485494.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000485494.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000492146.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000492146.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000492146.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000492146.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000497586.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000497586.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000497586.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000497586.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000497586.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000497586.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000497586.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | ENST00000497586.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013998.hg.1 | NM_016329.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03026656.hg.1 | ENST00000296295.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03026656.hg.1 | ENST00000358080.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03026656.hg.1 | ENST00000394750.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03026656.hg.1 | ENST00000394752.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03026656.hg.1 | NM_016329.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03026656.hg.1 | TC03001464.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0300208088.hg.1 | ENST00000394752.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300208088.hg.1 | ENST00000492146.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300208088.hg.1 | NM_016329.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300208088.hg.1 | ENST00000394752.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300208088.hg.1 | ENST00000492146.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300208088.hg.1 | NM_016329.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300208088.hg.1 | TC0300013998.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000394752.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000470575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000470575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000470575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000470575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000470575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000470575.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000474837.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000474837.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000474837.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000474837.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000474837.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000482396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000483069.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000483069.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000483069.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000483069.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000483069.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000483069.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000483069.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000485494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000485494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000485494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000492146.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000492146.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000492146.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000492146.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000497586.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000497586.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000497586.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000497586.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000497586.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000497586.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000497586.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | ENST00000497586.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013998.hg.1 | NM_016329.3 | Refseq |

SFMBT1
Genomic Map