Assay Details
| Gene Symbol: METTL14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020961.3 | NP_066012.1 | 3 - 4 | 409 | 136 | |
| XM_005263166.4 | XP_005263223.1 | 3 - 4 | 404 | 136 | ||
| XM_011532162.2 | XP_011530464.1 | 3 - 4 | 404 | 136 | ||
| GenBank mRNA | AB046847.1 | - | 3 - 4 | 364 | 136 | |
| AK055555.1 | - | 3 - 4 | 387 | 136 | ||
| BC006565.2 | - | 3 - 4 | 360 | 3010654 | 136 | |
| BC007449.2 | - | 3 - 4 | 360 | 3010654 | 136 | |
| BG773151.1 | - | 3 - 4 | 414 | 136 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04000608.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000502564.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000502564.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000502564.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000502564.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000502564.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000502564.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000502564.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000506780.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000506780.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000506780.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000506780.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000506780.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000508801.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000508801.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000508801.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000508801.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000508801.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000508801.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000626212.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000626212.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000626212.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000626212.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000626212.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000626212.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000626212.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04008749.hg.1 | ENST00000388822.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04008749.hg.1 | ENST00000506780.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04008749.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04008749.hg.1 | TC04000608.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04008750.hg.1 | ENST00000388822.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04008750.hg.1 | ENST00000506780.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04008750.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04008750.hg.1 | TC04000608.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000502564.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000502564.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000502564.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000502564.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000502564.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000502564.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000502564.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000506780.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000506780.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000506780.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000506780.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000506780.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000508801.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000508801.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000508801.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000508801.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000508801.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000508801.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000626212.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000626212.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000626212.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000626212.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000626212.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000626212.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000626212.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008547.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400158642.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400158642.hg.1 | ENST00000502564.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400158642.hg.1 | ENST00000626212.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400158642.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400158642.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400158642.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400158642.hg.1 | ENST00000502564.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400158642.hg.1 | ENST00000626212.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400158642.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400158642.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400158642.hg.1 | TC0400008547.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400158638.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400158638.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400158638.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400158638.hg.1 | ENST00000388822.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400158638.hg.1 | ENST00000628452.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400158638.hg.1 | NM_020961.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400158638.hg.1 | TC0400008547.hg.1 | Clariom D TC |

METTL14
Genomic Map