Assay Details
| Gene Symbol: SLC25A21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171170.1 | NP_001164641.1 | 5 - 6 | 640 | 64 | |
| NM_030631.3 | NP_085134.1 | 5 - 6 | 640 | 64 | ||
| XM_011537287.2 | XP_011535589.1 | 6 - 7 | 422 | 64 | ||
| XM_011537288.2 | XP_011535590.1 | 5 - 6 | 514 | 64 | ||
| XM_011537289.2 | XP_011535591.1 | 4 - 5 | 423 | 64 | ||
| GenBank mRNA | AJ278148.1 | - | 5 - 6 | 640 | 64 | |
| AK289575.1 | - | 5 - 6 | 403 | 64 | ||
| BC101521.1 | - | 5 - 6 | 376 | 8069027 | 64 | |
| BC113365.1 | - | 5 - 6 | 376 | 8322417 | 64 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14001051.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR14012019.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14012019.hg.1 | ENST00000555449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14012019.hg.1 | NM_001171170.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14012019.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14012019.hg.1 | TC14001051.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR14012018.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14012018.hg.1 | ENST00000555449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14012018.hg.1 | NM_001171170.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14012018.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14012018.hg.1 | TC14001051.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000546428.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000546428.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000546428.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000546428.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000546428.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000555449.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000555449.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000555449.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000555449.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000555449.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000555449.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000555449.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000555449.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000555449.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000555449.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000555449.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000555449.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000555449.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000556444.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000556444.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000557611.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000557611.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000557611.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000622765.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000622765.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000622765.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000622765.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000622765.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000622765.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000622765.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000622765.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000622765.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000622765.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000622765.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000622765.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000622765.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000622765.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | NM_001171170.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | NM_001171170.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | NM_001171170.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | NM_001171170.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | NM_001171170.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | NM_001171170.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | NM_001171170.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | NM_001171170.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | NM_001171170.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | NM_001171170.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | NM_001171170.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | NM_001171170.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | NM_001171170.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | NM_001171170.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | NM_001171170.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000546428.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000546428.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000546428.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000546428.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000546428.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000555449.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000555449.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000555449.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000555449.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000555449.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000555449.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000555449.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000555449.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000555449.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000555449.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000555449.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000555449.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000555449.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000556444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000556444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000557611.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000557611.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000557611.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000622765.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000622765.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000622765.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000622765.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000622765.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000622765.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000622765.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000622765.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000622765.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000622765.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000622765.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000622765.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000622765.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000622765.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | NM_001171170.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | NM_001171170.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | NM_001171170.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | NM_001171170.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | NM_001171170.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | NM_001171170.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | NM_001171170.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | NM_001171170.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | NM_001171170.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | NM_001171170.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | NM_001171170.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | NM_001171170.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | NM_001171170.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | NM_001171170.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | NM_001171170.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400166081.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166081.hg.1 | ENST00000555449.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166081.hg.1 | ENST00000556444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166081.hg.1 | ENST00000622765.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166081.hg.1 | NM_001171170.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400166081.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400166081.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166081.hg.1 | ENST00000555449.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166081.hg.1 | ENST00000556444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166081.hg.1 | ENST00000622765.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166081.hg.1 | NM_001171170.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400166081.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400166081.hg.1 | TC1400008973.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400166083.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166083.hg.1 | ENST00000546428.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166083.hg.1 | ENST00000555449.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166083.hg.1 | ENST00000556444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166083.hg.1 | ENST00000622765.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166083.hg.1 | NM_001171170.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400166083.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400166083.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166083.hg.1 | ENST00000546428.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166083.hg.1 | ENST00000555449.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166083.hg.1 | ENST00000556444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166083.hg.1 | ENST00000622765.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166083.hg.1 | NM_001171170.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400166083.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400166083.hg.1 | TC1400008973.hg.1 | Clariom D TC |

SLC25A21
Genomic Map