Assay Details
| Gene Symbol: SLC25A21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171170.1 | NP_001164641.1 | 8 - 9 | 1094 | 93 | |
| NM_030631.3 | NP_085134.1 | 8 - 9 | 1094 | 93 | ||
| XM_011537287.2 | XP_011535589.1 | 9 - 10 | 876 | 93 | ||
| XM_011537288.2 | XP_011535590.1 | 8 - 9 | 968 | 93 | ||
| XM_011537289.2 | XP_011535591.1 | 7 - 8 | 877 | 93 | ||
| GenBank mRNA | AJ278148.1 | - | 8 - 9 | 1094 | 93 | |
| AK289575.1 | - | 8 - 9 | 857 | 93 | ||
| BC101521.1 | - | 8 - 9 | 830 | 8069027 | 93 | |
| BC113365.1 | - | 8 - 9 | 830 | 8322417 | 93 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14001051.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR14012016.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14012016.hg.1 | ENST00000555449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14012016.hg.1 | NM_001171170.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14012016.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14012016.hg.1 | TC14001051.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR14012017.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14012017.hg.1 | ENST00000555449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14012017.hg.1 | NM_001171170.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14012017.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14012017.hg.1 | TC14001051.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000546428.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000546428.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000546428.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000546428.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000546428.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000555449.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000555449.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000555449.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000555449.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000555449.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000555449.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000555449.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000555449.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000555449.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000555449.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000555449.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000555449.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000555449.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000556444.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000556444.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000557611.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000557611.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000557611.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000622765.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000622765.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000622765.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000622765.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000622765.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000622765.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000622765.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000622765.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000622765.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000622765.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000622765.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000622765.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000622765.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | ENST00000622765.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | NM_001171170.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | NM_001171170.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | NM_001171170.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | NM_001171170.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | NM_001171170.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | NM_001171170.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | NM_001171170.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | NM_001171170.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | NM_001171170.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | NM_001171170.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | NM_001171170.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | NM_001171170.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | NM_001171170.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | NM_001171170.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | NM_001171170.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008973.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400166079.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166079.hg.1 | ENST00000555449.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166079.hg.1 | ENST00000622765.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166079.hg.1 | NM_001171170.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400166079.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400166079.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166079.hg.1 | ENST00000555449.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166079.hg.1 | ENST00000622765.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166079.hg.1 | NM_001171170.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400166079.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400166079.hg.1 | TC1400008973.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000546428.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000546428.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000546428.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000546428.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000546428.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000555449.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000555449.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000555449.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000555449.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000555449.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000555449.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000555449.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000555449.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000555449.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000555449.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000555449.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000555449.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000555449.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000556444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000556444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000557611.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000557611.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000557611.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000622765.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000622765.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000622765.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000622765.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000622765.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000622765.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000622765.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000622765.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000622765.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000622765.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000622765.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000622765.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000622765.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | ENST00000622765.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | NM_001171170.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | NM_001171170.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | NM_001171170.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | NM_001171170.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | NM_001171170.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | NM_001171170.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | NM_001171170.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | NM_001171170.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | NM_001171170.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | NM_001171170.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | NM_001171170.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | NM_001171170.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | NM_001171170.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | NM_001171170.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | NM_001171170.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008973.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400166078.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166078.hg.1 | ENST00000555449.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166078.hg.1 | ENST00000622765.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166078.hg.1 | NM_001171170.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400166078.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400166078.hg.1 | ENST00000331299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166078.hg.1 | ENST00000555449.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166078.hg.1 | ENST00000622765.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400166078.hg.1 | NM_001171170.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400166078.hg.1 | NM_030631.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400166078.hg.1 | TC1400008973.hg.1 | Clariom D TC |

SLC25A21
Genomic Map