| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Gene 1.0 ST Array |
XM_001716731_at |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
XM_001724506_at |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
XM_001716723_at |
- |
- |
| Affymetrix Human Gene 1.1 ST Array |
8055911 |
- |
- |
| Affymetrix HT HG-U133+ PM Array Plate |
224256_PM_at |
- |
- |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012856.hg.1 |
ENST00000321694.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012856.hg.1 |
ENST00000321694.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012856.hg.1 |
ENST00000321694.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012856.hg.1 |
ENST00000539956.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012856.hg.1 |
ENST00000539956.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012856.hg.1 |
ENST00000539956.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012856.hg.1 |
ENST00000539956.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012856.hg.1 |
ENST00000539956.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012856.hg.1 |
ENST00000539956.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012856.hg.1 |
ENST00000621133.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200012856.hg.1 |
ENST00000621133.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012856.hg.1 |
ENST00000321694.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012856.hg.1 |
ENST00000321694.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012856.hg.1 |
ENST00000321694.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012856.hg.1 |
ENST00000539956.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012856.hg.1 |
ENST00000539956.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012856.hg.1 |
ENST00000539956.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012856.hg.1 |
ENST00000539956.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012856.hg.1 |
ENST00000539956.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012856.hg.1 |
ENST00000539956.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012856.hg.1 |
ENST00000621133.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200012856.hg.1 |
ENST00000621133.1 |
Ensembl |
| Human Genome U133 Plus 2.0 Array |
224256_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
224256_PM_at |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
224256_at8 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
224256_at7 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
224256_at2 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
224256_at6 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
224256_at1 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
224256_at4 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
224256_at3 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
224256_at9 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
224256_at10 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
224256_at5 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
224256_at11 |
- |
- |
| Human Genome U133 Set |
224256_PM_at |
- |
- |
| Human Genome U133 Set |
224256_at |
- |
- |