| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008424.hg.1 |
ENST00000225371.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008424.hg.1 |
ENST00000225371.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008424.hg.1 |
ENST00000225371.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008424.hg.1 |
ENST00000225371.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008424.hg.1 |
ENST00000225371.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008424.hg.1 |
ENST00000225371.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008424.hg.1 |
ENST00000225371.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008424.hg.1 |
ENST00000225371.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008424.hg.1 |
ENST00000225371.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008424.hg.1 |
ENST00000225371.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008424.hg.1 |
ENST00000225371.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008424.hg.1 |
ENST00000225371.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008424.hg.1 |
ENST00000225371.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008424.hg.1 |
ENST00000225371.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008424.hg.1 |
NM_000502.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008424.hg.1 |
NM_000502.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008424.hg.1 |
NM_000502.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008424.hg.1 |
NM_000502.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008424.hg.1 |
NM_000502.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008424.hg.1 |
NM_000502.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008424.hg.1 |
NM_000502.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008424.hg.1 |
NM_000502.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008424.hg.1 |
NM_000502.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008424.hg.1 |
NM_000502.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008424.hg.1 |
NM_000502.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008424.hg.1 |
NM_000502.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008424.hg.1 |
NM_000502.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008424.hg.1 |
NM_000502.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17009258.hg.1 |
ENST00000225371.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17009258.hg.1 |
NM_000502.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17009258.hg.1 |
TC17000711.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1700162641.hg.1 |
ENST00000225371.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700162641.hg.1 |
NM_000502.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700162641.hg.1 |
TC1700008424.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700162641.hg.1 |
ENST00000225371.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700162641.hg.1 |
NM_000502.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008424.hg.1 |
ENST00000225371.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008424.hg.1 |
ENST00000225371.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008424.hg.1 |
ENST00000225371.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008424.hg.1 |
ENST00000225371.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008424.hg.1 |
ENST00000225371.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008424.hg.1 |
ENST00000225371.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008424.hg.1 |
ENST00000225371.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008424.hg.1 |
ENST00000225371.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008424.hg.1 |
ENST00000225371.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008424.hg.1 |
ENST00000225371.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008424.hg.1 |
ENST00000225371.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008424.hg.1 |
ENST00000225371.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008424.hg.1 |
ENST00000225371.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008424.hg.1 |
ENST00000225371.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008424.hg.1 |
NM_000502.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008424.hg.1 |
NM_000502.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008424.hg.1 |
NM_000502.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008424.hg.1 |
NM_000502.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008424.hg.1 |
NM_000502.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008424.hg.1 |
NM_000502.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008424.hg.1 |
NM_000502.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008424.hg.1 |
NM_000502.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008424.hg.1 |
NM_000502.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008424.hg.1 |
NM_000502.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008424.hg.1 |
NM_000502.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008424.hg.1 |
NM_000502.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008424.hg.1 |
NM_000502.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008424.hg.1 |
NM_000502.5 |
Refseq |