Assay Details
| Gene Symbol: MLLT10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195626.1 | NP_001182555.1 | 12 - 13 | 1699 | 101 | |
| NM_001324297.1 | NP_001311226.1 | 15 - 16 | 2532 | 101 | ||
| GenBank mRNA | AY598745.1 | - | 12 - 13 | 1699 | 101 | |
| BC129946.1 | - | 13 - 14 | 1742 | 101 | ||
| BX648210.1 | - | 6 - 7 | 784 | 101 | ||
More Information
Set Membership: |
|

MLLT10
Genomic Map