| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17010707.hg.1 |
ENST00000335666.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17010707.hg.1 |
NM_181790.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17010707.hg.1 |
TC17000829.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008848.hg.1 |
ENST00000335666.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008848.hg.1 |
ENST00000335666.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008848.hg.1 |
ENST00000335666.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008848.hg.1 |
ENST00000335666.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008848.hg.1 |
ENST00000335666.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008848.hg.1 |
ENST00000335666.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008848.hg.1 |
ENST00000582579.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008848.hg.1 |
ENST00000582579.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008848.hg.1 |
ENST00000582579.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008848.hg.1 |
ENST00000582579.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008848.hg.1 |
ENST00000582579.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008848.hg.1 |
ENST00000582579.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008848.hg.1 |
ENST00000582579.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008848.hg.1 |
ENST00000582579.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008848.hg.1 |
ENST00000582579.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008848.hg.1 |
ENST00000585308.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008848.hg.1 |
ENST00000585308.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008848.hg.1 |
ENST00000585308.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008848.hg.1 |
ENST00000585308.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008848.hg.1 |
ENST00000585308.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008848.hg.1 |
ENST00000585308.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008848.hg.1 |
NM_181790.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008848.hg.1 |
NM_181790.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008848.hg.1 |
NM_181790.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008848.hg.1 |
NM_181790.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008848.hg.1 |
NM_181790.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17010705.hg.1 |
ENST00000335666.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17010705.hg.1 |
NM_181790.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17010705.hg.1 |
TC17000829.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17010706.hg.1 |
ENST00000335666.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17010706.hg.1 |
NM_181790.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17010706.hg.1 |
TC17000829.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17010704.hg.1 |
ENST00000335666.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17010704.hg.1 |
NM_181790.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17010704.hg.1 |
TC17000829.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1700166070.hg.1 |
ENST00000335666.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700166070.hg.1 |
ENST00000582579.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700166070.hg.1 |
ENST00000585308.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700166070.hg.1 |
NM_181790.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700166070.hg.1 |
TC1700008848.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700166070.hg.1 |
ENST00000335666.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700166070.hg.1 |
ENST00000582579.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700166070.hg.1 |
ENST00000585308.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700166070.hg.1 |
NM_181790.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700166073.hg.1 |
ENST00000335666.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700166073.hg.1 |
ENST00000582579.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700166073.hg.1 |
ENST00000585308.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700166073.hg.1 |
NM_181790.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700166073.hg.1 |
TC1700008848.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700166073.hg.1 |
ENST00000335666.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700166073.hg.1 |
ENST00000582579.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700166073.hg.1 |
ENST00000585308.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700166073.hg.1 |
NM_181790.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700166074.hg.1 |
ENST00000335666.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700166074.hg.1 |
ENST00000582579.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700166074.hg.1 |
NM_181790.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700166074.hg.1 |
TC1700008848.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700166074.hg.1 |
ENST00000335666.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700166074.hg.1 |
ENST00000582579.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700166074.hg.1 |
NM_181790.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008848.hg.1 |
ENST00000335666.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008848.hg.1 |
ENST00000335666.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008848.hg.1 |
ENST00000335666.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008848.hg.1 |
ENST00000335666.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008848.hg.1 |
ENST00000335666.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008848.hg.1 |
ENST00000335666.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008848.hg.1 |
ENST00000582579.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008848.hg.1 |
ENST00000582579.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008848.hg.1 |
ENST00000582579.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008848.hg.1 |
ENST00000582579.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008848.hg.1 |
ENST00000582579.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008848.hg.1 |
ENST00000582579.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008848.hg.1 |
ENST00000582579.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008848.hg.1 |
ENST00000582579.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008848.hg.1 |
ENST00000582579.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008848.hg.1 |
ENST00000585308.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008848.hg.1 |
ENST00000585308.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008848.hg.1 |
ENST00000585308.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008848.hg.1 |
ENST00000585308.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008848.hg.1 |
ENST00000585308.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008848.hg.1 |
ENST00000585308.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008848.hg.1 |
NM_181790.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008848.hg.1 |
NM_181790.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008848.hg.1 |
NM_181790.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008848.hg.1 |
NM_181790.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700008848.hg.1 |
NM_181790.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700166068.hg.1 |
ENST00000335666.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700166068.hg.1 |
ENST00000582579.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700166068.hg.1 |
ENST00000585308.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700166068.hg.1 |
NM_181790.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700166068.hg.1 |
TC1700008848.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700166068.hg.1 |
ENST00000335666.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700166068.hg.1 |
ENST00000582579.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700166068.hg.1 |
ENST00000585308.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700166068.hg.1 |
NM_181790.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700166072.hg.1 |
ENST00000335666.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700166072.hg.1 |
ENST00000582579.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700166072.hg.1 |
ENST00000585308.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700166072.hg.1 |
NM_181790.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700166072.hg.1 |
TC1700008848.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700166072.hg.1 |
ENST00000335666.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700166072.hg.1 |
ENST00000582579.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700166072.hg.1 |
ENST00000585308.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700166072.hg.1 |
NM_181790.1 |
Refseq |