Assay Details
| Gene Symbol: CASKIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020764.3 | NP_065815.1 | 8 - 9 | 869 | 93 | |
| GenBank mRNA | AF451977.1 | - | 8 - 9 | 935 | 93 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16000777.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR16011431.hg.1 | ENST00000343516.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16011431.hg.1 | ENST00000382453.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16011431.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16011431.hg.1 | TC16000777.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000562055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000562055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000562055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000562055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000564289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000564289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000564289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600169184.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600169184.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600169184.hg.1 | TC1600009096.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600169184.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600169184.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000562055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000562055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000562055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000562055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000564289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000564289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000564289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | NM_020764.3 | Refseq |

CASKIN1
Genomic Map