Assay Details
| Gene Symbol: CASKIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020764.3 | NP_065815.1 | 10 - 11 | 1078 | 59 | |
| GenBank mRNA | AB037727.1 | - | 2 - 3 | 215 | 59 | |
| AF451977.1 | - | 10 - 11 | 1144 | 59 | ||
| BC042011.1 | - | 1 - 2 | 97 | 59 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16000777.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000562055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000562055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000562055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000562055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000564289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000564289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | ENST00000564289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16011429.hg.1 | ENST00000343516.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16011429.hg.1 | ENST00000382453.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16011429.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16011429.hg.1 | TC16000777.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000562055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000562055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000562055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000562055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000564289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000564289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | ENST00000564289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009096.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600169182.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600169182.hg.1 | NM_020764.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600169182.hg.1 | TC1600009096.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600169182.hg.1 | ENST00000343516.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600169182.hg.1 | NM_020764.3 | Refseq |

CASKIN1
Genomic Map