| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X014489.hg.1 |
ENST00000218328.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X014489.hg.1 |
ENST00000262854.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X014489.hg.1 |
ENST00000342160.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X014489.hg.1 |
ENST00000396323.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X014489.hg.1 |
NM_031407.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X014489.hg.1 |
TC0X001067.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0X00166808.hg.1 |
ENST00000218328.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00166808.hg.1 |
ENST00000262854.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00166808.hg.1 |
ENST00000342160.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00166808.hg.1 |
ENST00000612484.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00166808.hg.1 |
NM_031407.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00166808.hg.1 |
ENST00000218328.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00166808.hg.1 |
ENST00000262854.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00166808.hg.1 |
ENST00000342160.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00166808.hg.1 |
ENST00000612484.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00166808.hg.1 |
NM_031407.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00166808.hg.1 |
TC0X00009767.hg.1 |
Clariom D TC |