Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | hydroxylysine kinase | HYKK | AGPHD1 | -308.0 | Sense | Chr. 15 - 78507564 - 78537373 on NCBI Build GRCh38 |
| Downstream | proteasome subunit alpha 4 | PSMA4 |
HC9, HsT17706, PSC9 |
1.5kb | Sense | Chr. 15 - 78540405 - 78549220 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map