Assay Details
| Gene Symbol: FHAD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017000202.1 | XP_016855691.1 | 10 - 11 | 1578 | 71 | |
| GenBank mRNA | AK057963.1 | - | 9 - 10 | 1319 | 71 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100018175.hg.1 | ENST00000375995.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018175.hg.1 | ENST00000375995.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018175.hg.1 | ENST00000375995.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018175.hg.1 | ENST00000375995.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018175.hg.1 | ENST00000375995.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018175.hg.1 | ENST00000375995.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018175.hg.1 | ENST00000375996.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018175.hg.1 | ENST00000375996.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018175.hg.1 | ENST00000375996.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018175.hg.1 | ENST00000375996.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018175.hg.1 | ENST00000375996.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018175.hg.1 | ENST00000375996.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018175.hg.1 | ENST00000375996.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018175.hg.1 | ENST00000375996.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018175.hg.1 | ENST00000375996.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018175.hg.1 | ENST00000375996.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018175.hg.1 | ENST00000375996.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018175.hg.1 | ENST00000375996.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018175.hg.1 | ENST00000375997.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018175.hg.1 | ENST00000375997.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018175.hg.1 | ENST00000375997.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018175.hg.1 | ENST00000375997.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018175.hg.1 | ENST00000375997.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018175.hg.1 | ENST00000375997.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018175.hg.1 | ENST00000375997.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018175.hg.1 | ENST00000375997.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018175.hg.1 | ENST00000375997.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018175.hg.1 | ENST00000375997.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018175.hg.1 | ENST00000375997.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018175.hg.1 | ENST00000459961.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018175.hg.1 | ENST00000459961.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018175.hg.1 | ENST00000459961.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018175.hg.1 | ENST00000459961.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018175.hg.1 | ENST00000459961.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018175.hg.1 | ENST00000459961.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018175.hg.1 | ENST00000459961.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018175.hg.1 | ENST00000459961.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018175.hg.1 | ENST00000459961.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018175.hg.1 | ENST00000472131.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018175.hg.1 | ENST00000472131.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018175.hg.1 | ENST00000472131.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018175.hg.1 | ENST00000472131.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018175.hg.1 | ENST00000472131.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018175.hg.1 | ENST00000483694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018175.hg.1 | ENST00000483694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018175.hg.1 | ENST00000483694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018175.hg.1 | ENST00000483694.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018175.hg.1 | ENST00000524761.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018175.hg.1 | ENST00000524761.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018175.hg.1 | ENST00000524761.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018175.hg.1 | ENST00000524761.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018175.hg.1 | ENST00000375995.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018175.hg.1 | ENST00000375995.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018175.hg.1 | ENST00000375995.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018175.hg.1 | ENST00000375995.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018175.hg.1 | ENST00000375995.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018175.hg.1 | ENST00000375995.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018175.hg.1 | ENST00000375996.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018175.hg.1 | ENST00000375996.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018175.hg.1 | ENST00000375996.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018175.hg.1 | ENST00000375996.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018175.hg.1 | ENST00000375996.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018175.hg.1 | ENST00000375996.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018175.hg.1 | ENST00000375996.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018175.hg.1 | ENST00000375996.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018175.hg.1 | ENST00000375996.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018175.hg.1 | ENST00000375996.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018175.hg.1 | ENST00000375996.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018175.hg.1 | ENST00000375996.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018175.hg.1 | ENST00000375997.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018175.hg.1 | ENST00000375997.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018175.hg.1 | ENST00000375997.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018175.hg.1 | ENST00000375997.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018175.hg.1 | ENST00000375997.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018175.hg.1 | ENST00000375997.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018175.hg.1 | ENST00000375997.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018175.hg.1 | ENST00000375997.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018175.hg.1 | ENST00000375997.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018175.hg.1 | ENST00000375997.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018175.hg.1 | ENST00000375997.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018175.hg.1 | ENST00000459961.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018175.hg.1 | ENST00000459961.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018175.hg.1 | ENST00000459961.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018175.hg.1 | ENST00000459961.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018175.hg.1 | ENST00000459961.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018175.hg.1 | ENST00000459961.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018175.hg.1 | ENST00000459961.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018175.hg.1 | ENST00000459961.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018175.hg.1 | ENST00000459961.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018175.hg.1 | ENST00000472131.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018175.hg.1 | ENST00000472131.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018175.hg.1 | ENST00000472131.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018175.hg.1 | ENST00000472131.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018175.hg.1 | ENST00000472131.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018175.hg.1 | ENST00000483694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018175.hg.1 | ENST00000483694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018175.hg.1 | ENST00000483694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018175.hg.1 | ENST00000483694.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018175.hg.1 | ENST00000524761.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018175.hg.1 | ENST00000524761.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018175.hg.1 | ENST00000524761.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018175.hg.1 | ENST00000524761.5 | Ensembl |

FHAD1
Genomic Map