Assay Details
| Gene Symbol: SPTAN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195532.1 | NP_001182461.1 | 22 - 23 | 3299 | 63 | |
| XM_006717247.2 | XP_006717310.1 | 22 - 23 | 3206 | 63 | ||
| XM_006717251.2 | XP_006717314.1 | 22 - 23 | 3293 | 63 | ||
| XM_006717252.2 | XP_006717315.1 | 22 - 23 | 3206 | 63 | ||
| XM_006717253.1 | XP_006717316.1 | 22 - 23 | 3206 | 63 | ||
| XM_017015060.1 | XP_016870549.1 | 22 - 23 | 3206 | 63 | ||
| XM_017015061.1 | XP_016870550.1 | 22 - 23 | 3264 | 63 | ||
| GenBank mRNA | AB191262.1 | - | 22 - 23 | 3264 | 63 | |
| AK300386.1 | - | 22 - 23 | 3267 | 63 | ||
| AL110273.1 | - | 2 - 3 | 186 | 63 | ||
| BC053521.1 | - | 22 - 23 | 3248 | 5529391 | 63 | |
More Information
Set Membership: |
|

SPTAN1
Genomic Map