Assay Details
| Gene Symbol: SPTAN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195532.1 | NP_001182461.1 | 35 - 36 | 4842 | 72 | |
| NM_003127.3 | NP_003118.2 | 36 - 37 | 4902 | 72 | ||
| XM_006717246.1 | XP_006717309.1 | 36 - 37 | 4809 | 72 | ||
| XM_006717249.1 | XP_006717312.1 | 36 - 37 | 4809 | 72 | ||
| XM_006717253.1 | XP_006717316.1 | 35 - 36 | 4749 | 72 | ||
| GenBank mRNA | AB191262.1 | - | 35 - 36 | 4807 | 72 | |
| AB209860.1 | - | 36 - 37 | 4864 | 72 | ||
| AL110273.1 | - | 15 - 16 | 1729 | 72 | ||
| BC053521.1 | - | 35 - 36 | 4791 | 5529391 | 72 | |
| J05243.1 | - | 36 - 37 | 4862 | 72 | ||
More Information
Set Membership: |
|

SPTAN1
Genomic Map