Assay Details
| Gene Symbol: RASEF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152573.3 | NP_689786.2 | 7 - 8 | 1288 | 92 | |
| XM_005251731.3 | XP_005251788.1 | 7 - 8 | 1779 | 92 | ||
| GenBank mRNA | AK290986.1 | - | 7 - 8 | 1030 | 92 | |
| BC023566.1 | - | 7 - 8 | 1288 | 4138119 | 92 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09001276.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0900010565.hg.1 | ENST00000340717.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010565.hg.1 | ENST00000340717.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010565.hg.1 | ENST00000340717.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010565.hg.1 | ENST00000340717.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010565.hg.1 | ENST00000376447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010565.hg.1 | ENST00000376447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010565.hg.1 | ENST00000376447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010565.hg.1 | ENST00000376447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010565.hg.1 | ENST00000376447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010565.hg.1 | ENST00000376447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010565.hg.1 | ENST00000376447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010565.hg.1 | ENST00000376447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010565.hg.1 | ENST00000376447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010565.hg.1 | ENST00000376447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010565.hg.1 | ENST00000376447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010565.hg.1 | ENST00000376447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010565.hg.1 | ENST00000376447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010565.hg.1 | ENST00000376447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010565.hg.1 | ENST00000376447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010565.hg.1 | ENST00000376447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010565.hg.1 | ENST00000376447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010565.hg.1 | ENST00000376447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010565.hg.1 | ENST00000376447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010565.hg.1 | ENST00000376447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010565.hg.1 | ENST00000376447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010565.hg.1 | NM_152573.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010565.hg.1 | NM_152573.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010565.hg.1 | NM_152573.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010565.hg.1 | NM_152573.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010565.hg.1 | NM_152573.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010565.hg.1 | NM_152573.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010565.hg.1 | NM_152573.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010565.hg.1 | NM_152573.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010565.hg.1 | NM_152573.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010565.hg.1 | NM_152573.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010565.hg.1 | NM_152573.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010565.hg.1 | NM_152573.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010565.hg.1 | NM_152573.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010565.hg.1 | NM_152573.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010565.hg.1 | NM_152573.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010565.hg.1 | NM_152573.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010565.hg.1 | NM_152573.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010565.hg.1 | NM_152573.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010565.hg.1 | NM_152573.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010565.hg.1 | NM_152573.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010565.hg.1 | NM_152573.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09015892.hg.1 | ENST00000376447.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09015892.hg.1 | NM_152573.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09015892.hg.1 | TC09001276.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0900010565.hg.1 | ENST00000340717.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010565.hg.1 | ENST00000340717.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010565.hg.1 | ENST00000340717.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010565.hg.1 | ENST00000340717.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010565.hg.1 | ENST00000376447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010565.hg.1 | ENST00000376447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010565.hg.1 | ENST00000376447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010565.hg.1 | ENST00000376447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010565.hg.1 | ENST00000376447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010565.hg.1 | ENST00000376447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010565.hg.1 | ENST00000376447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010565.hg.1 | ENST00000376447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010565.hg.1 | ENST00000376447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010565.hg.1 | ENST00000376447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010565.hg.1 | ENST00000376447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010565.hg.1 | ENST00000376447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010565.hg.1 | ENST00000376447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010565.hg.1 | ENST00000376447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010565.hg.1 | ENST00000376447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010565.hg.1 | ENST00000376447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010565.hg.1 | ENST00000376447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010565.hg.1 | ENST00000376447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010565.hg.1 | ENST00000376447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010565.hg.1 | ENST00000376447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010565.hg.1 | ENST00000376447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010565.hg.1 | NM_152573.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010565.hg.1 | NM_152573.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010565.hg.1 | NM_152573.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010565.hg.1 | NM_152573.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010565.hg.1 | NM_152573.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010565.hg.1 | NM_152573.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010565.hg.1 | NM_152573.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010565.hg.1 | NM_152573.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010565.hg.1 | NM_152573.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010565.hg.1 | NM_152573.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010565.hg.1 | NM_152573.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010565.hg.1 | NM_152573.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010565.hg.1 | NM_152573.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010565.hg.1 | NM_152573.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010565.hg.1 | NM_152573.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010565.hg.1 | NM_152573.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010565.hg.1 | NM_152573.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010565.hg.1 | NM_152573.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010565.hg.1 | NM_152573.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010565.hg.1 | NM_152573.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010565.hg.1 | NM_152573.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900172032.hg.1 | ENST00000376447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900172032.hg.1 | NM_152573.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900172032.hg.1 | ENST00000376447.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900172032.hg.1 | NM_152573.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900172032.hg.1 | TC0900010565.hg.1 | Clariom D TC |

RASEF
Genomic Map