Assay Details
| Gene Symbol: KIF24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194313.2 | NP_919289.2 | 6 - 7 | 1333 | 67 | |
| XM_005251451.2 | XP_005251508.1 | 2 - 3 | 226 | 67 | ||
| XM_011517861.1 | XP_011516163.1 | 7 - 8 | 1554 | 67 | ||
| XM_011517863.2 | XP_011516165.1 | 6 - 7 | 2643 | 67 | ||
| XM_017014697.1 | XP_016870186.1 | 6 - 7 | 1647 | 67 | ||
| XM_017014698.1 | XP_016870187.1 | 5 - 6 | 1420 | 67 | ||
| GenBank mRNA | AK310014.1 | - | 7 - 8 | 1554 | 67 | |
| BP871643.1 | - | 3 - 4 | 387 | 67 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09001031.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000379166.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000379166.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000379166.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000379166.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000379166.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000379166.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000379166.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000379166.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000379166.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000379166.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000379166.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000379166.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000379166.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000379166.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000379166.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000379166.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000379174.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000379174.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000379174.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000379174.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000379174.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000379174.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000379174.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000379174.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000379174.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000379174.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000379174.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000379174.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000402558.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000402558.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000402558.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000402558.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000402558.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000402558.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000402558.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000402558.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000402558.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000402558.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000402558.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000402558.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000402558.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000402558.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000402558.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | ENST00000410494.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | NM_194313.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | NM_194313.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | NM_194313.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | NM_194313.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | NM_194313.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | NM_194313.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | NM_194313.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | NM_194313.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | NM_194313.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | NM_194313.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | NM_194313.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | NM_194313.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | NM_194313.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | NM_194313.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | NM_194313.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009892.hg.1 | NM_194313.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09013517.hg.1 | ENST00000345050.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09013517.hg.1 | ENST00000379166.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09013517.hg.1 | ENST00000379174.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09013517.hg.1 | ENST00000402558.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09013517.hg.1 | ENST00000420188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09013517.hg.1 | NM_194313.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09013517.hg.1 | TC09001031.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0900167775.hg.1 | ENST00000379166.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900167775.hg.1 | ENST00000379174.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900167775.hg.1 | ENST00000402558.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900167775.hg.1 | NM_194313.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900167775.hg.1 | ENST00000379166.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900167775.hg.1 | ENST00000379174.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900167775.hg.1 | ENST00000402558.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900167775.hg.1 | NM_194313.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900167775.hg.1 | TC0900009892.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000379166.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000379166.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000379166.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000379166.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000379166.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000379166.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000379166.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000379166.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000379166.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000379166.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000379166.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000379166.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000379166.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000379166.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000379166.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000379166.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000379174.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000379174.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000379174.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000379174.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000379174.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000379174.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000379174.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000379174.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000379174.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000379174.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000379174.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000379174.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000402558.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000402558.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000402558.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000402558.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000402558.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000402558.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000402558.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000402558.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000402558.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000402558.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000402558.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000402558.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000402558.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000402558.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000402558.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | ENST00000410494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | NM_194313.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | NM_194313.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | NM_194313.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | NM_194313.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | NM_194313.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | NM_194313.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | NM_194313.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | NM_194313.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | NM_194313.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | NM_194313.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | NM_194313.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | NM_194313.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | NM_194313.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | NM_194313.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | NM_194313.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009892.hg.1 | NM_194313.3 | Refseq |

KIF24
Genomic Map