| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04019908.hg.1 |
TC04001425.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04019908.hg.1 |
ENST00000394854.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04019908.hg.1 |
ENST00000507176.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04019908.hg.1 |
ENST00000512215.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04019908.hg.1 |
ENST00000523694.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04019908.hg.1 |
NM_000944.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400178333.hg.1 |
ENST00000394854.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400178333.hg.1 |
ENST00000507176.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400178333.hg.1 |
ENST00000512215.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400178333.hg.1 |
NM_000944.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400178333.hg.1 |
ENST00000394854.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400178333.hg.1 |
ENST00000507176.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400178333.hg.1 |
ENST00000512215.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400178333.hg.1 |
NM_000944.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400178333.hg.1 |
TC0400011440.hg.1 |
Clariom D TC |