Assay Details
| Gene Symbol: MRPS35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021821.3 | NP_068593.2 | 6 - 7 | 702 | 152 | |
| GenBank mRNA | AF068296.1 | - | 1 - 2 | 96 | 152 | |
| AF182422.1 | - | 6 - 7 | 678 | 152 | ||
| AK075515.1 | - | 6 - 7 | 640 | 152 | ||
| AL512733.1 | - | 5 - 6 | 495 | 152 | ||
| BC015862.1 | - | 6 - 7 | 647 | 4689849 | 152 | |
| BC028346.2 | - | 6 - 7 | 619 | 152 | ||
| BC063515.1 | - | 6 - 7 | 625 | 152 | ||
| BC109372.1 | - | 6 - 7 | 639 | 6715646 | 152 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12000270.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200007161.hg.1 | ENST00000081029.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007161.hg.1 | ENST00000081029.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007161.hg.1 | ENST00000081029.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007161.hg.1 | ENST00000081029.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007161.hg.1 | ENST00000081029.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007161.hg.1 | ENST00000081029.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007161.hg.1 | ENST00000081029.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007161.hg.1 | ENST00000081029.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007161.hg.1 | ENST00000081029.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007161.hg.1 | ENST00000081029.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007161.hg.1 | ENST00000081029.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007161.hg.1 | ENST00000081029.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007161.hg.1 | ENST00000081029.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007161.hg.1 | ENST00000536569.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007161.hg.1 | ENST00000536569.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007161.hg.1 | ENST00000536569.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007161.hg.1 | ENST00000536569.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007161.hg.1 | ENST00000536569.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007161.hg.1 | ENST00000536569.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007161.hg.1 | ENST00000538315.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007161.hg.1 | ENST00000538315.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007161.hg.1 | ENST00000538315.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007161.hg.1 | ENST00000538315.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007161.hg.1 | ENST00000538315.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007161.hg.1 | ENST00000538315.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007161.hg.1 | ENST00000538315.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007161.hg.1 | ENST00000538315.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007161.hg.1 | ENST00000538315.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007161.hg.1 | ENST00000538315.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007161.hg.1 | ENST00000542199.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007161.hg.1 | ENST00000542199.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007161.hg.1 | ENST00000542199.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007161.hg.1 | ENST00000542199.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007161.hg.1 | ENST00000542199.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007161.hg.1 | ENST00000542199.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007161.hg.1 | ENST00000542199.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007161.hg.1 | ENST00000542199.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007161.hg.1 | ENST00000542199.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007161.hg.1 | ENST00000542791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007161.hg.1 | ENST00000542791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007161.hg.1 | ENST00000542791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007161.hg.1 | ENST00000542791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007161.hg.1 | NM_001190864.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007161.hg.1 | NM_001190864.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007161.hg.1 | NM_001190864.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007161.hg.1 | NM_001190864.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007161.hg.1 | NM_001190864.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007161.hg.1 | NM_001190864.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007161.hg.1 | NM_001190864.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007161.hg.1 | NM_001190864.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007161.hg.1 | NM_001190864.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007161.hg.1 | NM_001190864.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007161.hg.1 | NM_001190864.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007161.hg.1 | NM_021821.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007161.hg.1 | NM_021821.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007161.hg.1 | NM_021821.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007161.hg.1 | NM_021821.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007161.hg.1 | NM_021821.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007161.hg.1 | NM_021821.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007161.hg.1 | NM_021821.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007161.hg.1 | NM_021821.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007161.hg.1 | NM_021821.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007161.hg.1 | NM_021821.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007161.hg.1 | NM_021821.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007161.hg.1 | NM_021821.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007161.hg.1 | NM_021821.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003303.hg.1 | ENST00000081029.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003303.hg.1 | ENST00000321446.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003303.hg.1 | ENST00000538315.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003303.hg.1 | NM_001190864.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003303.hg.1 | NM_021821.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003303.hg.1 | TC12000270.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003301.hg.1 | ENST00000081029.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003301.hg.1 | NM_021821.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003301.hg.1 | TC12000270.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1200150307.hg.1 | ENST00000081029.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200150307.hg.1 | ENST00000538315.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200150307.hg.1 | ENST00000542199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200150307.hg.1 | NM_001190864.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200150307.hg.1 | NM_021821.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200150307.hg.1 | ENST00000081029.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200150307.hg.1 | ENST00000538315.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200150307.hg.1 | ENST00000542199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200150307.hg.1 | NM_001190864.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200150307.hg.1 | NM_021821.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200150307.hg.1 | TC1200007161.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200007161.hg.1 | ENST00000081029.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007161.hg.1 | ENST00000081029.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007161.hg.1 | ENST00000081029.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007161.hg.1 | ENST00000081029.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007161.hg.1 | ENST00000081029.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007161.hg.1 | ENST00000081029.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007161.hg.1 | ENST00000081029.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007161.hg.1 | ENST00000081029.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007161.hg.1 | ENST00000081029.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007161.hg.1 | ENST00000081029.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007161.hg.1 | ENST00000081029.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007161.hg.1 | ENST00000081029.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007161.hg.1 | ENST00000081029.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007161.hg.1 | ENST00000536569.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007161.hg.1 | ENST00000536569.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007161.hg.1 | ENST00000536569.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007161.hg.1 | ENST00000536569.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007161.hg.1 | ENST00000536569.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007161.hg.1 | ENST00000536569.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007161.hg.1 | ENST00000538315.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007161.hg.1 | ENST00000538315.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007161.hg.1 | ENST00000538315.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007161.hg.1 | ENST00000538315.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007161.hg.1 | ENST00000538315.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007161.hg.1 | ENST00000538315.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007161.hg.1 | ENST00000538315.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007161.hg.1 | ENST00000538315.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007161.hg.1 | ENST00000538315.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007161.hg.1 | ENST00000538315.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007161.hg.1 | ENST00000542199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007161.hg.1 | ENST00000542199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007161.hg.1 | ENST00000542199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007161.hg.1 | ENST00000542199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007161.hg.1 | ENST00000542199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007161.hg.1 | ENST00000542199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007161.hg.1 | ENST00000542199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007161.hg.1 | ENST00000542199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007161.hg.1 | ENST00000542199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007161.hg.1 | ENST00000542791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007161.hg.1 | ENST00000542791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007161.hg.1 | ENST00000542791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007161.hg.1 | ENST00000542791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007161.hg.1 | NM_001190864.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007161.hg.1 | NM_001190864.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007161.hg.1 | NM_001190864.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007161.hg.1 | NM_001190864.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007161.hg.1 | NM_001190864.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007161.hg.1 | NM_001190864.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007161.hg.1 | NM_001190864.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007161.hg.1 | NM_001190864.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007161.hg.1 | NM_001190864.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007161.hg.1 | NM_001190864.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007161.hg.1 | NM_001190864.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007161.hg.1 | NM_021821.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007161.hg.1 | NM_021821.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007161.hg.1 | NM_021821.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007161.hg.1 | NM_021821.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007161.hg.1 | NM_021821.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007161.hg.1 | NM_021821.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007161.hg.1 | NM_021821.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007161.hg.1 | NM_021821.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007161.hg.1 | NM_021821.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007161.hg.1 | NM_021821.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007161.hg.1 | NM_021821.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007161.hg.1 | NM_021821.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007161.hg.1 | NM_021821.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200150305.hg.1 | ENST00000081029.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200150305.hg.1 | ENST00000536569.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200150305.hg.1 | ENST00000542199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200150305.hg.1 | NM_021821.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200150305.hg.1 | ENST00000081029.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200150305.hg.1 | ENST00000536569.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200150305.hg.1 | ENST00000542199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200150305.hg.1 | NM_021821.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200150305.hg.1 | TC1200007161.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200150306.hg.1 | ENST00000081029.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200150306.hg.1 | ENST00000542199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200150306.hg.1 | NM_021821.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200150306.hg.1 | ENST00000081029.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200150306.hg.1 | ENST00000542199.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200150306.hg.1 | NM_021821.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200150306.hg.1 | TC1200007161.hg.1 | Clariom D TC |

MRPS35
Genomic Map