Assay Details
| Gene Symbol: UAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001324113.1 | NP_001311042.1 | 9 - 10 | 1632 | 140 | |
| NM_001324114.1 | NP_001311043.1 | 8 - 9 | 1520 | 140 | ||
| NM_001324115.1 | NP_001311044.1 | 9 - 10 | 1859 | 140 | ||
| NM_003115.4 | NP_003106.3 | 8 - 9 | 1681 | 140 | ||
| GenBank mRNA | AB011004.1 | - | 7 - 8 | 1352 | 140 | |
| AK312685.1 | - | 8 - 9 | 1607 | 140 | ||
| BC009377.2 | - | 8 - 9 | 1654 | 4121439 | 140 | |
| S73498.1 | - | 8 - 9 | 1584 | 140 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01001422.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000474728.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000474728.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000474728.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000476240.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000476240.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000486089.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000486089.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000486089.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | ENST00000486089.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010381.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022227.hg.1 | ENST00000271469.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022227.hg.1 | ENST00000367924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022227.hg.1 | ENST00000367925.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022227.hg.1 | ENST00000367926.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022227.hg.1 | ENST00000486089.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022227.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01022227.hg.1 | TC01001422.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000367926.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000474728.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000474728.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000474728.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000476240.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000476240.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000486089.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000486089.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000486089.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | ENST00000486089.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010381.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100176506.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176506.hg.1 | ENST00000367924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176506.hg.1 | ENST00000367925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176506.hg.1 | ENST00000367926.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176506.hg.1 | ENST00000486089.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176506.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100176506.hg.1 | ENST00000271469.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176506.hg.1 | ENST00000367924.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176506.hg.1 | ENST00000367925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176506.hg.1 | ENST00000367926.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176506.hg.1 | ENST00000486089.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100176506.hg.1 | NM_003115.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100176506.hg.1 | TC0100010381.hg.1 | Clariom D TC |

UAP1
Genomic Map