Assay Details
| Gene Symbol: NCR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145457.2 | NP_001138929.2 | 2 - 3 | 106 | 66 | |
| NM_001145458.2 | NP_001138930.2 | 2 - 3 | 106 | 66 | ||
| NM_004829.6 | NP_004820.2 | 2 - 3 | 106 | 66 | ||
| XM_011527528.2 | XP_011525830.1 | 2 - 3 | 214 | 66 | ||
| XM_011527529.2 | XP_011525831.1 | 2 - 3 | 210 | 66 | ||
| XM_011527530.2 | XP_011525832.1 | 2 - 3 | 202 | 66 | ||
| GenBank mRNA | AJ001383.1 | - | 2 - 3 | 107 | 66 | |
| AJ006121.1 | - | 2 - 3 | 75 | 66 | ||
| BC064806.1 | - | 2 - 3 | 93 | 5218848 | 66 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19000886.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000291890.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000291890.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000291890.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000291890.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000291890.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000291890.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000291890.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000291890.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000291890.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000291890.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000291890.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000338835.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000338835.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000338835.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000338835.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000338835.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000338835.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000338835.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000338835.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000338835.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000350790.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000350790.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000350790.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000350790.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000350790.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000350790.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000350790.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000350790.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000357397.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000357397.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000357397.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000357397.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000357397.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000357397.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000598576.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000598576.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000598576.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000598576.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000598576.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000598576.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000598576.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000598576.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000598576.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000601137.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000601137.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000601137.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000601137.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000601137.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000601137.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000601903.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000601903.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000601903.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000601903.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19012275.hg.1 | ENST00000291890.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19012275.hg.1 | ENST00000338835.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19012275.hg.1 | ENST00000447255.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19012275.hg.1 | NM_001145457.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19012275.hg.1 | NM_001145458.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19012275.hg.1 | NM_004829.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19012275.hg.1 | TC19000886.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1900169509.hg.1 | ENST00000291890.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900169509.hg.1 | ENST00000338835.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900169509.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900169509.hg.1 | ENST00000598576.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900169509.hg.1 | ENST00000601137.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900169509.hg.1 | ENST00000601903.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900169509.hg.1 | TC1900008882.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900169509.hg.1 | ENST00000291890.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900169509.hg.1 | ENST00000338835.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900169509.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900169509.hg.1 | ENST00000598576.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900169509.hg.1 | ENST00000601137.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900169509.hg.1 | ENST00000601903.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000291890.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000291890.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000291890.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000291890.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000291890.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000291890.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000291890.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000291890.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000291890.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000291890.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000291890.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000338835.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000338835.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000338835.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000338835.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000338835.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000338835.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000338835.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000338835.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000338835.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000350790.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000350790.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000350790.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000350790.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000350790.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000350790.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000350790.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000350790.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000357397.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000357397.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000357397.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000357397.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000357397.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000357397.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000598576.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000598576.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000598576.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000598576.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000598576.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000598576.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000598576.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000598576.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000598576.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000601137.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000601137.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000601137.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000601137.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000601137.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000601137.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000601903.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000601903.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000601903.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000601903.1 | Ensembl |

NCR1
Genomic Map