Assay Details
| Gene Symbol: NCR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001242356.2 | NP_001229285.1 | 4 - 5 | 438 | 71 | |
| NM_004829.6 | NP_004820.2 | 5 - 6 | 723 | 71 | ||
| GenBank mRNA | AJ001383.1 | - | 5 - 6 | 724 | 71 | |
| AJ006122.1 | - | 4 - 5 | 407 | 71 | ||
| AY346373.1 | - | 3 - 4 | 364 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000291890.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000291890.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000291890.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000291890.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000291890.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000291890.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000291890.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000291890.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000291890.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000291890.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000291890.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000338835.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000338835.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000338835.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000338835.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000338835.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000338835.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000338835.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000338835.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000338835.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000350790.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000350790.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000350790.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000350790.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000350790.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000350790.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000350790.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000350790.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000357397.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000357397.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000357397.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000357397.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000357397.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000357397.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000598576.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000598576.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000598576.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000598576.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000598576.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000598576.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000598576.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000598576.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000598576.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000601137.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000601137.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000601137.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000601137.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000601137.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000601137.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000601903.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000601903.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000601903.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008882.hg.1 | ENST00000601903.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19012279.hg.1 | ENST00000291890.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19012279.hg.1 | ENST00000350790.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19012279.hg.1 | ENST00000357397.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19012279.hg.1 | ENST00000447255.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19012279.hg.1 | NM_001145457.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19012279.hg.1 | NM_001242356.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19012279.hg.1 | NM_004829.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19012279.hg.1 | TC19000886.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19012277.hg.1 | ENST00000291890.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19012277.hg.1 | ENST00000338835.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19012277.hg.1 | ENST00000350790.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19012277.hg.1 | ENST00000357397.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19012277.hg.1 | ENST00000447255.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19012277.hg.1 | NM_001145457.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19012277.hg.1 | NM_001145458.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19012277.hg.1 | NM_001242356.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19012277.hg.1 | NM_001242357.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19012277.hg.1 | NM_004829.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19012277.hg.1 | TC19000886.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000291890.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000291890.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000291890.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000291890.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000291890.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000291890.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000291890.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000291890.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000291890.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000291890.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000291890.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000338835.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000338835.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000338835.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000338835.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000338835.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000338835.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000338835.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000338835.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000338835.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000350790.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000350790.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000350790.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000350790.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000350790.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000350790.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000350790.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000350790.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000357397.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000357397.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000357397.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000357397.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000357397.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000357397.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000594765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000598576.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000598576.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000598576.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000598576.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000598576.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000598576.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000598576.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000598576.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000598576.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000601137.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000601137.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000601137.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000601137.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000601137.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000601137.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000601903.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000601903.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000601903.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008882.hg.1 | ENST00000601903.1 | Ensembl |

NCR1
Genomic Map