Assay Details
| Gene Symbol: UBE2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282161.1 | NP_001269090.1 | 4 - 5 | 481 | 154 | |
| NM_003336.3 | NP_003327.2 | 4 - 5 | 488 | 154 | ||
| GenBank mRNA | AK223045.1 | - | 4 - 5 | 390 | 154 | |
| AK223064.1 | - | 4 - 5 | 387 | 154 | ||
| AK301675.1 | - | 3 - 4 | 399 | 154 | ||
| AK313092.1 | - | 4 - 5 | 392 | 154 | ||
| BC010175.2 | - | 4 - 5 | 381 | 3051041 | 154 | |
| DA068506.1 | - | 4 - 5 | 385 | 154 | ||
| DA599154.1 | - | 4 - 5 | 488 | 154 | ||
More Information
Set Membership: |
|

UBE2A
Genomic Map