| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007102.hg.1 |
ENST00000375471.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007102.hg.1 |
ENST00000375471.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007102.hg.1 |
ENST00000375471.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007102.hg.1 |
ENST00000375471.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007102.hg.1 |
ENST00000375471.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007102.hg.1 |
ENST00000375471.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007102.hg.1 |
ENST00000375471.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007102.hg.1 |
ENST00000375471.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007102.hg.1 |
ENST00000375471.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007102.hg.1 |
ENST00000375471.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007102.hg.1 |
ENST00000375471.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007102.hg.1 |
ENST00000375471.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007102.hg.1 |
ENST00000375471.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007102.hg.1 |
ENST00000375471.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007102.hg.1 |
ENST00000375471.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007102.hg.1 |
ENST00000375471.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007102.hg.1 |
ENST00000375471.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007102.hg.1 |
ENST00000375471.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007102.hg.1 |
ENST00000375471.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007102.hg.1 |
ENST00000460293.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007102.hg.1 |
ENST00000460293.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007102.hg.1 |
ENST00000460293.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007102.hg.1 |
ENST00000460293.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007102.hg.1 |
ENST00000460293.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007102.hg.1 |
ENST00000460293.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007102.hg.1 |
ENST00000460293.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007102.hg.1 |
ENST00000483501.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007102.hg.1 |
ENST00000483501.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007102.hg.1 |
ENST00000483501.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007102.hg.1 |
ENST00000483501.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007102.hg.1 |
ENST00000483501.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007102.hg.1 |
NM_013358.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007102.hg.1 |
NM_013358.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007102.hg.1 |
NM_013358.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007102.hg.1 |
NM_013358.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007102.hg.1 |
NM_013358.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007102.hg.1 |
NM_013358.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007102.hg.1 |
NM_013358.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007102.hg.1 |
NM_013358.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007102.hg.1 |
NM_013358.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007102.hg.1 |
NM_013358.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007102.hg.1 |
NM_013358.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007102.hg.1 |
NM_013358.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007102.hg.1 |
NM_013358.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007102.hg.1 |
NM_013358.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007102.hg.1 |
NM_013358.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007102.hg.1 |
NM_013358.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007102.hg.1 |
NM_013358.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007102.hg.1 |
NM_013358.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007102.hg.1 |
NM_013358.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01003652.hg.1 |
ENST00000375471.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01003652.hg.1 |
ENST00000413717.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01003652.hg.1 |
ENST00000460293.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01003652.hg.1 |
ENST00000536552.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01003652.hg.1 |
ENST00000537499.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01003652.hg.1 |
NM_013358.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01003652.hg.1 |
TC01000230.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0100149546.hg.1 |
ENST00000375471.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100149546.hg.1 |
ENST00000460293.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100149546.hg.1 |
NM_013358.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100149546.hg.1 |
ENST00000375471.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100149546.hg.1 |
ENST00000460293.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100149546.hg.1 |
NM_013358.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100149546.hg.1 |
TC0100007102.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100007102.hg.1 |
ENST00000375471.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007102.hg.1 |
ENST00000375471.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007102.hg.1 |
ENST00000375471.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007102.hg.1 |
ENST00000375471.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007102.hg.1 |
ENST00000375471.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007102.hg.1 |
ENST00000375471.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007102.hg.1 |
ENST00000375471.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007102.hg.1 |
ENST00000375471.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007102.hg.1 |
ENST00000375471.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007102.hg.1 |
ENST00000375471.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007102.hg.1 |
ENST00000375471.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007102.hg.1 |
ENST00000375471.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007102.hg.1 |
ENST00000375471.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007102.hg.1 |
ENST00000375471.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007102.hg.1 |
ENST00000375471.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007102.hg.1 |
ENST00000375471.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007102.hg.1 |
ENST00000375471.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007102.hg.1 |
ENST00000375471.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007102.hg.1 |
ENST00000375471.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007102.hg.1 |
ENST00000460293.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007102.hg.1 |
ENST00000460293.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007102.hg.1 |
ENST00000460293.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007102.hg.1 |
ENST00000460293.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007102.hg.1 |
ENST00000460293.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007102.hg.1 |
ENST00000460293.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007102.hg.1 |
ENST00000460293.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007102.hg.1 |
ENST00000483501.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007102.hg.1 |
ENST00000483501.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007102.hg.1 |
ENST00000483501.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007102.hg.1 |
ENST00000483501.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007102.hg.1 |
ENST00000483501.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007102.hg.1 |
NM_013358.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007102.hg.1 |
NM_013358.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007102.hg.1 |
NM_013358.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007102.hg.1 |
NM_013358.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007102.hg.1 |
NM_013358.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007102.hg.1 |
NM_013358.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007102.hg.1 |
NM_013358.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007102.hg.1 |
NM_013358.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007102.hg.1 |
NM_013358.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007102.hg.1 |
NM_013358.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007102.hg.1 |
NM_013358.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007102.hg.1 |
NM_013358.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007102.hg.1 |
NM_013358.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007102.hg.1 |
NM_013358.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007102.hg.1 |
NM_013358.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007102.hg.1 |
NM_013358.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007102.hg.1 |
NM_013358.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007102.hg.1 |
NM_013358.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007102.hg.1 |
NM_013358.2 |
Refseq |