Assay Details
| Gene Symbol: EXT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000127.2 | NP_000118.2 | 4 - 5 | 2059 | 122 | |
| GenBank mRNA | AK313129.1 | - | 4 - 5 | 1807 | 122 | |
| BC001174.1 | - | 4 - 5 | 1452 | 3502232 | 122 | |
| S79639.1 | - | 4 - 5 | 1937 | 122 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08001548.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
skeletal system development
ossification
glycosaminoglycan biosynthetic process
protein glycosylation
signal transduction
gastrulation
axon guidance
endoderm development
mesoderm development
heparan sulfate proteoglycan biosynthetic process
heparan sulfate proteoglycan biosynthetic process, polysaccharide chain biosynthetic process
olfactory bulb development
cellular polysaccharide biosynthetic process
embryonic skeletal joint development
acetylglucosaminyltransferase activity
glucuronosyltransferase activity
transferase activity, transferring glycosyl groups
heparan sulfate N-acetylglucosaminyltransferase activity
protein homodimerization activity
metal ion binding
protein heterodimerization activity
glucuronosyl-N-acetylglucosaminyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferase activity
N-acetylglucosaminyl-proteoglycan 4-beta-glucuronosyltransferase activity
skeletal system development
ossification
glycosaminoglycan biosynthetic process
protein glycosylation
signal transduction
gastrulation
axon guidance
endoderm development
mesoderm development
heparan sulfate proteoglycan biosynthetic process
heparan sulfate proteoglycan biosynthetic process, polysaccharide chain biosynthetic process
olfactory bulb development
cellular polysaccharide biosynthetic process
embryonic skeletal joint development
acetylglucosaminyltransferase activity
glucuronosyltransferase activity
transferase activity, transferring glycosyl groups
heparan sulfate N-acetylglucosaminyltransferase activity
protein homodimerization activity
metal ion binding
protein heterodimerization activity
glucuronosyl-N-acetylglucosaminyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferase activity
N-acetylglucosaminyl-proteoglycan 4-beta-glucuronosyltransferase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800011579.hg.1 | ENST00000378204.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011579.hg.1 | ENST00000378204.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011579.hg.1 | ENST00000378204.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011579.hg.1 | ENST00000378204.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011579.hg.1 | ENST00000378204.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011579.hg.1 | ENST00000378204.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011579.hg.1 | ENST00000378204.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011579.hg.1 | ENST00000378204.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011579.hg.1 | ENST00000378204.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011579.hg.1 | ENST00000378204.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011579.hg.1 | ENST00000378204.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011579.hg.1 | ENST00000378204.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011579.hg.1 | ENST00000378204.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011579.hg.1 | ENST00000378204.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011579.hg.1 | ENST00000378204.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011579.hg.1 | ENST00000378204.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011579.hg.1 | ENST00000378204.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011579.hg.1 | ENST00000378204.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011579.hg.1 | ENST00000378204.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011579.hg.1 | ENST00000378204.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011579.hg.1 | ENST00000436216.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011579.hg.1 | ENST00000436216.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011579.hg.1 | ENST00000436216.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011579.hg.1 | ENST00000436216.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011579.hg.1 | ENST00000436216.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011579.hg.1 | ENST00000437196.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011579.hg.1 | ENST00000437196.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011579.hg.1 | ENST00000437196.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011579.hg.1 | ENST00000437196.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011579.hg.1 | ENST00000437196.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011579.hg.1 | ENST00000437196.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011579.hg.1 | ENST00000437196.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011579.hg.1 | ENST00000437196.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011579.hg.1 | ENST00000437196.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011579.hg.1 | ENST00000437196.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011579.hg.1 | ENST00000437196.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011579.hg.1 | ENST00000437196.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011579.hg.1 | NM_000127.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011579.hg.1 | NM_000127.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011579.hg.1 | NM_000127.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011579.hg.1 | NM_000127.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011579.hg.1 | NM_000127.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011579.hg.1 | NM_000127.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011579.hg.1 | NM_000127.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011579.hg.1 | NM_000127.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011579.hg.1 | NM_000127.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011579.hg.1 | NM_000127.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011579.hg.1 | NM_000127.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011579.hg.1 | NM_000127.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011579.hg.1 | NM_000127.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011579.hg.1 | NM_000127.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011579.hg.1 | NM_000127.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011579.hg.1 | NM_000127.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011579.hg.1 | NM_000127.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011579.hg.1 | NM_000127.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08020398.hg.1 | ENST00000378204.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08020398.hg.1 | NM_000127.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08020398.hg.1 | TC08001548.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0800011579.hg.1 | ENST00000378204.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011579.hg.1 | ENST00000378204.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011579.hg.1 | ENST00000378204.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011579.hg.1 | ENST00000378204.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011579.hg.1 | ENST00000378204.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011579.hg.1 | ENST00000378204.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011579.hg.1 | ENST00000378204.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011579.hg.1 | ENST00000378204.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011579.hg.1 | ENST00000378204.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011579.hg.1 | ENST00000378204.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011579.hg.1 | ENST00000378204.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011579.hg.1 | ENST00000378204.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011579.hg.1 | ENST00000378204.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011579.hg.1 | ENST00000378204.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011579.hg.1 | ENST00000378204.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011579.hg.1 | ENST00000378204.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011579.hg.1 | ENST00000378204.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011579.hg.1 | ENST00000378204.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011579.hg.1 | ENST00000378204.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011579.hg.1 | ENST00000378204.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011579.hg.1 | ENST00000436216.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011579.hg.1 | ENST00000436216.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011579.hg.1 | ENST00000436216.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011579.hg.1 | ENST00000436216.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011579.hg.1 | ENST00000436216.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011579.hg.1 | ENST00000437196.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011579.hg.1 | ENST00000437196.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011579.hg.1 | ENST00000437196.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011579.hg.1 | ENST00000437196.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011579.hg.1 | ENST00000437196.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011579.hg.1 | ENST00000437196.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011579.hg.1 | ENST00000437196.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011579.hg.1 | ENST00000437196.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011579.hg.1 | ENST00000437196.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011579.hg.1 | ENST00000437196.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011579.hg.1 | ENST00000437196.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011579.hg.1 | ENST00000437196.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011579.hg.1 | NM_000127.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011579.hg.1 | NM_000127.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011579.hg.1 | NM_000127.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011579.hg.1 | NM_000127.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011579.hg.1 | NM_000127.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011579.hg.1 | NM_000127.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011579.hg.1 | NM_000127.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011579.hg.1 | NM_000127.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011579.hg.1 | NM_000127.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011579.hg.1 | NM_000127.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011579.hg.1 | NM_000127.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011579.hg.1 | NM_000127.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011579.hg.1 | NM_000127.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011579.hg.1 | NM_000127.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011579.hg.1 | NM_000127.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011579.hg.1 | NM_000127.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011579.hg.1 | NM_000127.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011579.hg.1 | NM_000127.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800178679.hg.1 | ENST00000378204.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800178679.hg.1 | ENST00000437196.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800178679.hg.1 | NM_000127.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800178679.hg.1 | ENST00000378204.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800178679.hg.1 | ENST00000437196.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800178679.hg.1 | NM_000127.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800178679.hg.1 | TC0800011579.hg.1 | Clariom D TC |

EXT1
Genomic Map