| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007181.hg.1 |
ENST00000238831.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007181.hg.1 |
ENST00000238831.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007181.hg.1 |
ENST00000238831.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007181.hg.1 |
ENST00000238831.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007181.hg.1 |
ENST00000238831.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007181.hg.1 |
ENST00000238831.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007181.hg.1 |
ENST00000238831.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007181.hg.1 |
ENST00000238831.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007181.hg.1 |
ENST00000238831.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007181.hg.1 |
ENST00000238831.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007181.hg.1 |
ENST00000238831.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007181.hg.1 |
ENST00000238831.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007181.hg.1 |
ENST00000238831.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007181.hg.1 |
ENST00000238831.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007181.hg.1 |
ENST00000238831.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007181.hg.1 |
ENST00000238831.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007181.hg.1 |
ENST00000238831.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007181.hg.1 |
ENST00000238831.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007181.hg.1 |
ENST00000238831.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007181.hg.1 |
ENST00000437765.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007181.hg.1 |
ENST00000437765.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007181.hg.1 |
ENST00000437765.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007181.hg.1 |
ENST00000437765.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007181.hg.1 |
ENST00000437765.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007181.hg.1 |
ENST00000437765.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007181.hg.1 |
ENST00000441084.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007181.hg.1 |
ENST00000441084.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007181.hg.1 |
ENST00000495355.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007181.hg.1 |
ENST00000495355.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007181.hg.1 |
ENST00000495355.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007181.hg.1 |
ENST00000495355.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007181.hg.1 |
NM_032312.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007181.hg.1 |
NM_032312.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007181.hg.1 |
NM_032312.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007181.hg.1 |
NM_032312.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007181.hg.1 |
NM_032312.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007181.hg.1 |
NM_032312.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007181.hg.1 |
NM_032312.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007181.hg.1 |
NM_032312.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007181.hg.1 |
NM_032312.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007181.hg.1 |
NM_032312.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007181.hg.1 |
NM_032312.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007181.hg.1 |
NM_032312.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02003055.hg.1 |
ENST00000238831.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02003055.hg.1 |
NM_032312.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02003055.hg.1 |
TC02000212.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0200007181.hg.1 |
ENST00000238831.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007181.hg.1 |
ENST00000238831.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007181.hg.1 |
ENST00000238831.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007181.hg.1 |
ENST00000238831.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007181.hg.1 |
ENST00000238831.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007181.hg.1 |
ENST00000238831.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007181.hg.1 |
ENST00000238831.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007181.hg.1 |
ENST00000238831.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007181.hg.1 |
ENST00000238831.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007181.hg.1 |
ENST00000238831.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007181.hg.1 |
ENST00000238831.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007181.hg.1 |
ENST00000238831.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007181.hg.1 |
ENST00000238831.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007181.hg.1 |
ENST00000238831.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007181.hg.1 |
ENST00000238831.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007181.hg.1 |
ENST00000238831.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007181.hg.1 |
ENST00000238831.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007181.hg.1 |
ENST00000238831.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007181.hg.1 |
ENST00000238831.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007181.hg.1 |
ENST00000437765.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007181.hg.1 |
ENST00000437765.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007181.hg.1 |
ENST00000437765.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007181.hg.1 |
ENST00000437765.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007181.hg.1 |
ENST00000437765.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007181.hg.1 |
ENST00000437765.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007181.hg.1 |
ENST00000441084.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007181.hg.1 |
ENST00000441084.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007181.hg.1 |
ENST00000495355.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007181.hg.1 |
ENST00000495355.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007181.hg.1 |
ENST00000495355.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007181.hg.1 |
ENST00000495355.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007181.hg.1 |
NM_032312.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007181.hg.1 |
NM_032312.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007181.hg.1 |
NM_032312.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007181.hg.1 |
NM_032312.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007181.hg.1 |
NM_032312.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007181.hg.1 |
NM_032312.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007181.hg.1 |
NM_032312.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007181.hg.1 |
NM_032312.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007181.hg.1 |
NM_032312.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007181.hg.1 |
NM_032312.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007181.hg.1 |
NM_032312.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007181.hg.1 |
NM_032312.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200149505.hg.1 |
ENST00000238831.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200149505.hg.1 |
ENST00000437765.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200149505.hg.1 |
NM_032312.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200149505.hg.1 |
ENST00000238831.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200149505.hg.1 |
ENST00000437765.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200149505.hg.1 |
NM_032312.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200149505.hg.1 |
TC0200007181.hg.1 |
Clariom D TC |