Assay Details
| Gene Symbol: CNST | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001139459.1 | NP_001132931.1 | 2 - 3 | 884 | 153 | |
| NM_152609.2 | NP_689822.2 | 2 - 3 | 758 | 153 | ||
| XM_005273081.3 | XP_005273138.2 | 2 - 3 | 543 | 153 | ||
| XM_011544110.2 | XP_011542412.1 | 2 - 3 | 609 | 153 | ||
| XM_011544111.1 | XP_011542413.1 | 2 - 3 | 780 | 153 | ||
| XM_017000488.1 | XP_016855977.1 | 2 - 3 | 591 | 153 | ||
| XM_017000489.1 | XP_016855978.1 | 2 - 3 | 658 | 153 | ||
| GenBank mRNA | AK056563.1 | - | 2 - 3 | 591 | 153 | |
| AK074142.1 | - | 2 - 3 | 521 | 153 | ||
| AL834246.2 | - | 2 - 3 | 592 | 153 | ||
| BC010228.2 | - | 2 - 3 | 760 | 4151228 | 153 | |
| BC036200.1 | - | 2 - 3 | 651 | 153 | ||
| BX647629.1 | - | 2 - 3 | 543 | 153 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01002000.hg
Additional Information:
This assay is in the 5'UTR region of the XM_017000488.1,XM_017000489.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01030945.hg.1 | ENST00000366512.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01030945.hg.1 | ENST00000366513.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01030945.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01030945.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01030945.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01030945.hg.1 | TC01002000.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366511.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366511.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366511.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366511.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366511.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01030943.hg.1 | ENST00000366512.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01030943.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01030943.hg.1 | TC01002000.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0100190259.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100190259.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100190259.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100190259.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100190259.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100190259.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100190259.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100190259.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100190259.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100190259.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100190259.hg.1 | TC0100012272.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366511.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366511.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366511.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366511.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366511.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100190254.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100190254.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100190254.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100190254.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100190254.hg.1 | TC0100012272.hg.1 | Clariom D TC |

CNST
Genomic Map