Assay Details
| Gene Symbol: CNST | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001139459.1 | NP_001132931.1 | 5 - 6 | 1207 | 81 | |
| NM_152609.2 | NP_689822.2 | 5 - 6 | 1081 | 81 | ||
| XM_005273081.3 | XP_005273138.2 | 5 - 6 | 866 | 81 | ||
| XM_005273083.4 | XP_005273140.1 | 4 - 5 | 784 | 81 | ||
| XM_011544110.2 | XP_011542412.1 | 5 - 6 | 932 | 81 | ||
| XM_011544111.1 | XP_011542413.1 | 5 - 6 | 1103 | 81 | ||
| XM_011544112.1 | XP_011542414.1 | 4 - 5 | 650 | 81 | ||
| XM_011544113.1 | XP_011542415.1 | 4 - 5 | 775 | 81 | ||
| XM_011544114.2 | XP_011542416.1 | 4 - 5 | 490 | 81 | ||
| XM_017000487.1 | XP_016855976.1 | 6 - 7 | 1121 | 81 | ||
| XM_017000488.1 | XP_016855977.1 | 5 - 6 | 864 | 81 | ||
| XM_017000489.1 | XP_016855978.1 | 5 - 6 | 931 | 81 | ||
| GenBank mRNA | AK056563.1 | - | 5 - 6 | 914 | 81 | |
| AL834246.2 | - | 5 - 6 | 915 | 81 | ||
| BC010228.2 | - | 5 - 6 | 1083 | 4151228 | 81 | |
| BX647629.1 | - | 5 - 6 | 866 | 81 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01002000.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01030948.hg.1 | ENST00000366512.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01030948.hg.1 | ENST00000366513.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01030948.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01030948.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01030948.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01030948.hg.1 | TC01002000.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366511.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366511.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366511.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366511.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366511.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01030950.hg.1 | ENST00000366512.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01030950.hg.1 | ENST00000366513.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01030950.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01030950.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01030950.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01030950.hg.1 | TC01002000.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0100190266.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100190266.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100190266.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100190266.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100190266.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100190266.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100190266.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100190266.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100190266.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100190266.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100190266.hg.1 | TC0100012272.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366511.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366511.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366511.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366511.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366511.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012272.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100190264.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100190264.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100190264.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100190264.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100190264.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100190264.hg.1 | ENST00000366512.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100190264.hg.1 | ENST00000366513.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100190264.hg.1 | ENST00000483271.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100190264.hg.1 | NM_001139459.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100190264.hg.1 | NM_152609.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100190264.hg.1 | TC0100012272.hg.1 | Clariom D TC |

CNST
Genomic Map