Assay Details
| Gene Symbol: NCR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001242356.2 | NP_001229285.1 | 2 - 3 | 107 | 85 | |
| NM_001242357.2 | NP_001229286.1 | 2 - 3 | 107 | 85 | ||
| GenBank mRNA | AJ006122.1 | - | 2 - 3 | 76 | 85 | |
| AJ006123.1 | - | 2 - 3 | 76 | 85 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Fc fragment of IgA receptor | FCAR |
CD89, CTB-61M7.2, FcalphaRI |
-14.9kb | Sense | Chr. 19 - 54874093 - 54891420 on NCBI Build GRCh38 |
| Downstream | NLR family pyrin domain containing 7 | NLRP7 |
CLR19.4, HYDM, NALP7, NOD12, PAN7, PYPAF3 |
14.0kb | Anti-sense | Chr. 19 - 54923509 - 54965184 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

NCR1
Genomic Map