| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000204637.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000204637.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000204637.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000204637.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000204637.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000204637.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000204637.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000204637.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000204637.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000204637.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000204637.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000204637.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000204637.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000593422.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000593422.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000593422.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000593422.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000593422.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000593422.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000593422.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000594009.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000594009.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000594009.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000594009.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000594009.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000594009.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000594009.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000594009.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000594009.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000594009.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000594009.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000594009.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000594009.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000595510.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000595510.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000595510.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000595510.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000595510.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000595510.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000595510.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000595510.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000595510.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000595510.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000595510.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000596435.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000596435.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000596435.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000596435.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000596435.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000596435.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000596435.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000596435.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000596435.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000596435.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000596435.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000596435.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000596435.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000597551.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000597551.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000597551.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000597551.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000597551.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000597551.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000597551.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000597551.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000597551.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000597551.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000597551.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000597551.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000597914.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000597914.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000597914.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000597914.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000598472.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000598472.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000598472.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000598472.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000598472.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000598472.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000598555.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000598555.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000598555.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000598555.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000598555.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000598555.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000598555.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000600084.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000600084.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000600084.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000600084.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000600084.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000600084.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000600084.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000600084.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000600084.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000600084.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000600084.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000600084.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000600084.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000600084.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000600429.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000600429.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000600429.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000600429.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000600429.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000600429.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000600429.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000600429.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000600429.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000600429.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000600429.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000600429.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000600429.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000601800.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000601800.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000601800.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000601800.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000601800.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000601800.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000601800.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000601800.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000601800.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000601800.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000601800.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000601800.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
ENST00000601800.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
NM_001204502.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
NM_001204502.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
NM_001204502.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
NM_001204502.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
NM_001204502.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
NM_001204502.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
NM_001204502.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
NM_001204502.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
NM_001204502.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
NM_001204502.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
NM_001204502.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
NM_001204502.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
NM_001204502.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
NM_001204502.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
NM_001204503.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
NM_001204503.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
NM_001204503.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
NM_001204503.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
NM_001204503.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
NM_001204503.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
NM_001204503.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
NM_001204503.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
NM_001204503.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
NM_001204503.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
NM_001204503.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
NM_001204503.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
NM_001204503.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
NM_001278637.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
NM_001278637.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
NM_001278637.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
NM_001278637.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
NM_001278637.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
NM_001278637.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
NM_001278637.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
NM_001278637.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
NM_001278637.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
NM_001278637.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
NM_001278637.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
NM_001278637.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
NM_001278637.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
NM_001278637.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
NM_001278638.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
NM_001278638.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
NM_001278638.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
NM_001278638.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
NM_001278638.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
NM_001278638.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
NM_001278638.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
NM_001278638.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
NM_001278638.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
NM_001278638.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
NM_001278638.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
NM_001278638.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
NM_001278638.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
NM_001459.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
NM_001459.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
NM_001459.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
NM_001459.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
NM_001459.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
NM_001459.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
NM_001459.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
NM_001459.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
NM_001459.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
NM_001459.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
NM_001459.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
NM_001459.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011779.hg.1 |
NM_001459.3 |
Refseq |