Assay Details
| Gene Symbol: PLPP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003711.3 | NP_003702.2 | 2 - 3 | 663 | 116 | |
| NR_103485.1 | 3 - 4 | 818 | 116 | |||
| GenBank mRNA | AB000888.1 | - | 2 - 3 | 261 | 116 | |
| AF014402.1 | - | 2 - 3 | 555 | 116 | ||
| AF017116.1 | - | 2 - 3 | 315 | 116 | ||
| BC039847.1 | - | 2 - 3 | 527 | 5762574 | 116 | |
| BC117133.1 | - | 2 - 3 | 264 | 40125684 | 116 | |
| BC143281.1 | - | 2 - 3 | 264 | 9051784 | 116 | |
| BC168254.1 | - | 3 - 4 | 419 | 116 | ||
More Information
Updated Mapping Information
Set Membership: |
|

PLPP1
Genomic Map