Assay Details
| Gene Symbol: PLPP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003711.3 | NP_003702.2 | 4 - 5 | 999 | 76 | |
| NM_176895.2 | NP_795714.1 | 4 - 5 | 1002 | 76 | ||
| NR_103485.1 | 5 - 6 | 1154 | 76 | |||
| XM_006714724.2 | XP_006714787.1 | 3 - 4 | 1139 | 76 | ||
| GenBank mRNA | AB000888.1 | - | 4 - 5 | 597 | 76 | |
| AF014402.1 | - | 4 - 5 | 891 | 76 | ||
| AF014403.1 | - | 4 - 5 | 894 | 76 | ||
| AF017116.1 | - | 4 - 5 | 651 | 76 | ||
| BC039847.1 | - | 4 - 5 | 863 | 5762574 | 76 | |
| BC117133.1 | - | 4 - 5 | 600 | 40125684 | 76 | |
| BC143281.1 | - | 4 - 5 | 600 | 9051784 | 76 | |
| BC168254.1 | - | 5 - 6 | 755 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
protein dephosphorylation
lipid metabolic process
phospholipid metabolic process
protein kinase C-activating G-protein coupled receptor signaling pathway
negative regulation of cell proliferation
germ cell migration
regulation of lipid metabolic process
sphingolipid biosynthetic process
intracellular steroid hormone receptor signaling pathway
androgen receptor signaling pathway
phospholipid dephosphorylation
phosphatidate phosphatase activity
sphingosine-1-phosphate phosphatase activity
lipid phosphatase activity
protein dephosphorylation
lipid metabolic process
phospholipid metabolic process
protein kinase C-activating G-protein coupled receptor signaling pathway
negative regulation of cell proliferation
germ cell migration
regulation of lipid metabolic process
sphingolipid biosynthetic process
intracellular steroid hormone receptor signaling pathway
androgen receptor signaling pathway
phospholipid dephosphorylation
phosphatidate phosphatase activity
sphingosine-1-phosphate phosphatase activity
lipid phosphatase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | ENST00000264775.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | ENST00000264775.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | ENST00000264775.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | ENST00000264775.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | ENST00000264775.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | ENST00000264775.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | ENST00000264775.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | ENST00000264775.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | ENST00000264775.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | ENST00000264775.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | ENST00000264775.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | ENST00000307259.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | ENST00000307259.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | ENST00000307259.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | ENST00000307259.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | ENST00000307259.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | ENST00000307259.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | ENST00000307259.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | ENST00000307259.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | ENST00000307259.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | ENST00000307259.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | ENST00000307259.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | ENST00000502765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | ENST00000502765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | ENST00000502765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | ENST00000502907.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | ENST00000502907.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | ENST00000502907.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | ENST00000502907.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | ENST00000509667.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | ENST00000509667.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | ENST00000509667.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | ENST00000509667.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | ENST00000509667.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | ENST00000509667.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | ENST00000515132.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | ENST00000515132.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | ENST00000515132.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | ENST00000515132.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | ENST00000520263.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | ENST00000520263.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | ENST00000520263.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | NM_003711.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | NM_003711.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | NM_003711.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | NM_003711.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | NM_003711.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | NM_003711.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | NM_003711.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | NM_003711.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | NM_003711.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | NM_003711.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | NM_003711.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | NM_003711.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | NM_003711.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | NM_176895.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | NM_176895.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | NM_176895.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | NM_176895.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | NM_176895.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | NM_176895.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | NM_176895.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | NM_176895.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | NM_176895.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | NM_176895.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | NM_176895.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | NM_176895.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | NM_176895.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | NR_001575.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | NR_001575.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | NR_001575.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | NR_001575.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | NR_001575.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | NR_103485.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | NR_103485.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | NR_103485.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | NR_103485.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | NR_103485.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | NR_103485.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | NR_103485.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | NR_103485.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | NR_103485.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | NR_103485.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | NR_103485.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | NR_103485.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | NR_103485.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010766.hg.1 | NR_103485.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | ENST00000264775.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | ENST00000264775.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | ENST00000264775.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | ENST00000264775.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | ENST00000264775.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | ENST00000264775.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | ENST00000264775.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | ENST00000264775.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | ENST00000264775.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | ENST00000264775.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | ENST00000264775.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | ENST00000307259.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | ENST00000307259.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | ENST00000307259.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | ENST00000307259.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | ENST00000307259.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | ENST00000307259.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | ENST00000307259.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | ENST00000307259.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | ENST00000307259.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | ENST00000307259.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | ENST00000307259.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | ENST00000502765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | ENST00000502765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | ENST00000502765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | ENST00000502907.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | ENST00000502907.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | ENST00000502907.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | ENST00000502907.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | ENST00000509667.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | ENST00000509667.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | ENST00000509667.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | ENST00000509667.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | ENST00000509667.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | ENST00000509667.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | ENST00000515132.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | ENST00000515132.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | ENST00000515132.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | ENST00000515132.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | ENST00000520263.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | ENST00000520263.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | ENST00000520263.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | NM_003711.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | NM_003711.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | NM_003711.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | NM_003711.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | NM_003711.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | NM_003711.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | NM_003711.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | NM_003711.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | NM_003711.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | NM_003711.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | NM_003711.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | NM_003711.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | NM_003711.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | NM_176895.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | NM_176895.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | NM_176895.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | NM_176895.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | NM_176895.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | NM_176895.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | NM_176895.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | NM_176895.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | NM_176895.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | NM_176895.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | NM_176895.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | NM_176895.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | NM_176895.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | NR_001575.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | NR_001575.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | NR_001575.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | NR_001575.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | NR_001575.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | NR_103485.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | NR_103485.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | NR_103485.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | NR_103485.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | NR_103485.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | NR_103485.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | NR_103485.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | NR_103485.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | NR_103485.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | NR_103485.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | NR_103485.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | NR_103485.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | NR_103485.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010766.hg.1 | NR_103485.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500175116.hg.1 | ENST00000264775.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500175116.hg.1 | ENST00000307259.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500175116.hg.1 | NM_003711.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500175116.hg.1 | NM_176895.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500175116.hg.1 | NR_103485.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500175116.hg.1 | ENST00000264775.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500175116.hg.1 | ENST00000307259.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500175116.hg.1 | NM_003711.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500175116.hg.1 | NM_176895.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500175116.hg.1 | NR_103485.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500175116.hg.1 | TC0500010766.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500175117.hg.1 | ENST00000264775.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500175117.hg.1 | ENST00000307259.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500175117.hg.1 | NM_003711.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500175117.hg.1 | NM_176895.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500175117.hg.1 | NR_103485.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500175117.hg.1 | ENST00000264775.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500175117.hg.1 | ENST00000307259.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500175117.hg.1 | NM_003711.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500175117.hg.1 | NM_176895.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500175117.hg.1 | NR_103485.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500175117.hg.1 | TC0500010766.hg.1 | Clariom D TC |

PLPP1
Genomic Map