Assay Details
| Gene Symbol: SYNJ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160302.1 | NP_001153774.1 | 27 - 28 | 3594 | 70 | |
| NM_003895.3 | NP_003886.3 | 28 - 29 | 3634 | 70 | ||
| NM_203446.2 | NP_982271.2 | 28 - 29 | 3634 | 70 | ||
| XM_017028494.1 | XP_016883983.1 | 27 - 28 | 3576 | 70 | ||
| XM_017028495.1 | XP_016883984.1 | 27 - 28 | 3634 | 70 | ||
| XM_017028496.1 | XP_016883985.1 | 26 - 27 | 3537 | 70 | ||
| XM_017028497.1 | XP_016883986.1 | 27 - 28 | 3674 | 70 | ||
| XM_017028500.1 | XP_016883989.1 | 28 - 29 | 3624 | 70 | ||
| XM_017028501.1 | XP_016883990.1 | 28 - 29 | 3624 | 70 | ||
| XM_017028502.1 | XP_016883991.1 | 27 - 28 | 3576 | 70 | ||
| XM_017028503.1 | XP_016883992.1 | 26 - 27 | 3537 | 70 | ||
| GenBank mRNA | AB020717.2 | - | 26 - 27 | 3530 | 70 | |
| AF009039.1 | - | 28 - 29 | 3560 | 70 | ||
| AF009040.1 | - | 28 - 29 | 3560 | 70 | ||
| AK295706.1 | - | 5 - 6 | 575 | 70 | ||
More Information
Set Membership: |
|

SYNJ1
Genomic Map