Assay Details
| Gene Symbol: UBE2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181762.2 | NP_861427.1 | 2 - 3 | 369 | 79 | |
| GenBank mRNA | AK297696.1 | - | 2 - 3 | 271 | 79 | |
| BI093289.1 | - | 2 - 3 | 159 | 79 | ||
More Information
Set Membership: |
|

UBE2A
Genomic Map