Assay Details
| Gene Symbol: SETD9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_136555.1 | 1 - 2 | 478 | 115 | ||
| NR_136556.1 | 1 - 2 | 478 | 115 | |||
| NR_136558.1 | 1 - 2 | 478 | 115 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000285947.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000285947.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000285947.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000285947.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000285947.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000285947.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000285947.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000285947.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000285947.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000285947.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000285947.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000285947.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000285947.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000285947.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000418299.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000418299.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000418299.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000418299.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000418299.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000418299.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000418299.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000418299.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000418299.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000423328.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000423328.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000463805.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000463805.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000463805.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000463805.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000463805.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000463805.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000472636.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000472636.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000472636.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000472636.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000475908.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000475908.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000475908.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000475908.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000475908.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000475908.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000475908.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000475908.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000475908.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000477359.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000477359.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000477359.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000477359.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000480414.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000480414.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000480414.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000480414.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000498322.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000498322.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000498322.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000498322.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000498322.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000624733.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000624733.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000624733.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000624733.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000628593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000628593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000628593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000628593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000628593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000628593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000628593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000628593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000628593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | ENST00000628593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | NM_001171990.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | NM_001171990.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | NM_001171990.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | NM_001171990.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | NM_001171990.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | NM_001171990.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | NM_001171990.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | NM_001171990.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | NM_001171990.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | NM_001171990.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | NM_001171990.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | NM_153706.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | NM_153706.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | NM_153706.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | NM_153706.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | NM_153706.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | NM_153706.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | NM_153706.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | NM_153706.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | NM_153706.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | NM_153706.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | NM_153706.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | NM_153706.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | NM_153706.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | NM_153706.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007457.hg.1 | NM_153706.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000285947.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000285947.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000285947.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000285947.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000285947.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000285947.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000285947.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000285947.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000285947.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000285947.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000285947.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000285947.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000285947.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000285947.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000418299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000418299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000418299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000418299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000418299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000418299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000418299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000418299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000418299.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000423328.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000423328.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000463805.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000463805.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000463805.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000463805.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000463805.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000463805.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000472636.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000472636.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000472636.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000472636.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000475908.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000475908.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000475908.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000475908.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000475908.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000475908.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000475908.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000475908.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000475908.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000477359.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000477359.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000477359.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000477359.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000480414.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000480414.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000480414.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000480414.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000498322.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000498322.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000498322.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000498322.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000498322.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000624733.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000624733.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000624733.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000624733.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000628593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000628593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000628593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000628593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000628593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000628593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000628593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000628593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000628593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | ENST00000628593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | NM_001171990.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | NM_001171990.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | NM_001171990.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | NM_001171990.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | NM_001171990.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | NM_001171990.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | NM_001171990.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | NM_001171990.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | NM_001171990.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | NM_001171990.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | NM_001171990.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | NM_153706.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | NM_153706.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | NM_153706.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | NM_153706.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | NM_153706.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | NM_153706.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | NM_153706.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | NM_153706.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | NM_153706.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | NM_153706.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | NM_153706.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | NM_153706.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | NM_153706.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | NM_153706.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007457.hg.1 | NM_153706.3 | Refseq |

SETD9
Genomic Map