Assay Details
| Gene Symbol: VSIG10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019086.5 | NP_061959.2 | 3 - 4 | 995 | 99 | |
| XM_011538502.1 | XP_011536804.1 | 3 - 4 | 995 | 99 | ||
| XM_017019506.1 | XP_016874995.1 | 3 - 4 | 1129 | 99 | ||
| GenBank mRNA | AK000681.1 | - | 3 - 4 | 1025 | 99 | |
| BC028698.2 | - | 3 - 4 | 939 | 4838234 | 99 | |
| BC034471.2 | - | 3 - 4 | 934 | 4825667 | 99 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12002017.hg.1
Additional Information:
This assay is in the 5'UTR region of the XM_017019506.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR12026505.hg.1 | ENST00000359236.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12026505.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12026505.hg.1 | TC12002017.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12026504.hg.1 | ENST00000359236.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12026504.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12026504.hg.1 | TC12002017.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000536905.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000536905.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000536905.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000536905.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000536905.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000536905.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000536905.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000536905.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000536905.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000538357.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000538357.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000538357.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000538357.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000538357.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000538357.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000540117.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000540117.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000541855.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000541855.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000541855.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000541855.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000541855.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000542195.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000542195.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000542195.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000542195.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000542195.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000542195.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000542195.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000545259.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000545259.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000545259.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000545259.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000545722.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000545722.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000612851.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000612851.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000612851.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000612851.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000612851.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000612851.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000612851.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000612851.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000536905.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000536905.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000536905.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000536905.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000536905.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000536905.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000536905.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000536905.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000536905.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000538357.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000538357.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000538357.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000538357.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000538357.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000538357.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000540117.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000540117.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000541855.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000541855.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000541855.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000541855.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000541855.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000542195.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000542195.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000542195.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000542195.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000542195.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000542195.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000542195.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000545259.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000545259.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000545259.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000545259.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000545722.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000545722.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000612851.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000612851.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000612851.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000612851.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000612851.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000612851.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000612851.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000612851.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200204556.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200204556.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200204556.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200204556.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200204556.hg.1 | TC1200012855.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200204555.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200204555.hg.1 | ENST00000538357.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200204555.hg.1 | ENST00000545722.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200204555.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200204555.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200204555.hg.1 | ENST00000538357.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200204555.hg.1 | ENST00000545722.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200204555.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200204555.hg.1 | TC1200012855.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200204557.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200204557.hg.1 | ENST00000545722.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200204557.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200204557.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200204557.hg.1 | ENST00000545722.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200204557.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200204557.hg.1 | TC1200012855.hg.1 | Clariom D TC |

VSIG10
Genomic Map