Assay Details
| Gene Symbol: VSIG10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019086.5 | NP_061959.2 | 5 - 6 | 1551 | 95 | |
| XM_005253908.4 | XP_005253965.1 | 4 - 5 | 904 | 95 | ||
| XM_006719467.2 | XP_006719530.1 | 4 - 5 | 1248 | 95 | ||
| XM_006719468.2 | XP_006719531.1 | 3 - 4 | 643 | 95 | ||
| XM_011538502.1 | XP_011536804.1 | 5 - 6 | 1551 | 95 | ||
| XM_017019504.1 | XP_016874993.1 | 6 - 7 | 1565 | 95 | ||
| XM_017019505.1 | XP_016874994.1 | 5 - 6 | 1171 | 95 | ||
| XM_017019506.1 | XP_016874995.1 | 5 - 6 | 1685 | 95 | ||
| GenBank mRNA | AK000681.1 | - | 5 - 6 | 1581 | 95 | |
| BC028698.2 | - | 5 - 6 | 1495 | 4838234 | 95 | |
| BC034471.2 | - | 5 - 6 | 1490 | 4825667 | 95 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12002017.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR12026502.hg.1 | ENST00000359236.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12026502.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12026502.hg.1 | TC12002017.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000536905.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000536905.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000536905.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000536905.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000536905.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000536905.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000536905.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000536905.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000536905.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000538357.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000538357.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000538357.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000538357.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000538357.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000538357.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000540117.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000540117.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000541855.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000541855.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000541855.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000541855.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000541855.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000542195.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000542195.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000542195.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000542195.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000542195.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000542195.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000542195.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000545259.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000545259.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000545259.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000545259.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000545722.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000545722.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000612851.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000612851.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000612851.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000612851.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000612851.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000612851.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000612851.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | ENST00000612851.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200204551.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200204551.hg.1 | ENST00000545259.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200204551.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200204551.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200204551.hg.1 | ENST00000545259.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200204551.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200204551.hg.1 | TC1200012855.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000536905.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000536905.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000536905.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000536905.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000536905.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000536905.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000536905.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000536905.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000536905.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000538357.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000538357.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000538357.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000538357.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000538357.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000538357.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000540117.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000540117.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000541855.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000541855.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000541855.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000541855.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000541855.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000542195.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000542195.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000542195.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000542195.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000542195.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000542195.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000542195.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000545259.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000545259.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000545259.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000545259.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000545722.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000545722.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000612851.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000612851.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000612851.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000612851.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000612851.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000612851.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000612851.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | ENST00000612851.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012855.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200204549.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200204549.hg.1 | ENST00000545259.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200204549.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200204549.hg.1 | ENST00000359236.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200204549.hg.1 | ENST00000545259.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200204549.hg.1 | NM_019086.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200204549.hg.1 | TC1200012855.hg.1 | Clariom D TC |

VSIG10
Genomic Map