| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012377.hg.1 |
ENST00000379102.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012377.hg.1 |
ENST00000379102.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012377.hg.1 |
ENST00000379102.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012377.hg.1 |
ENST00000379102.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012377.hg.1 |
ENST00000379102.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012377.hg.1 |
ENST00000379103.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012377.hg.1 |
ENST00000379103.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012377.hg.1 |
ENST00000379103.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012377.hg.1 |
ENST00000379103.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012377.hg.1 |
ENST00000379103.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012377.hg.1 |
ENST00000379103.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012377.hg.1 |
ENST00000379103.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012377.hg.1 |
ENST00000379103.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012377.hg.1 |
ENST00000379103.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012377.hg.1 |
ENST00000460380.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012377.hg.1 |
ENST00000460380.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012377.hg.1 |
ENST00000460380.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012377.hg.1 |
ENST00000460380.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012377.hg.1 |
ENST00000460380.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012377.hg.1 |
ENST00000460380.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012377.hg.1 |
ENST00000460380.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012377.hg.1 |
ENST00000460380.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012377.hg.1 |
ENST00000460380.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012377.hg.1 |
ENST00000460380.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012377.hg.1 |
ENST00000503642.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012377.hg.1 |
ENST00000503642.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012377.hg.1 |
ENST00000503642.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012377.hg.1 |
ENST00000503642.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012377.hg.1 |
ENST00000503642.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012377.hg.1 |
ENST00000508816.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012377.hg.1 |
ENST00000508816.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012377.hg.1 |
ENST00000508816.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012377.hg.1 |
ENST00000508862.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012377.hg.1 |
ENST00000508862.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012377.hg.1 |
ENST00000508862.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012377.hg.1 |
ENST00000508862.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012377.hg.1 |
ENST00000508862.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012377.hg.1 |
ENST00000508862.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012377.hg.1 |
ENST00000508862.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012377.hg.1 |
ENST00000582343.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012377.hg.1 |
ENST00000582343.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012377.hg.1 |
ENST00000582343.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012377.hg.1 |
ENST00000582343.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012377.hg.1 |
ENST00000582343.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012377.hg.1 |
ENST00000582343.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012377.hg.1 |
ENST00000582343.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012377.hg.1 |
NM_001076680.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012377.hg.1 |
NM_001076680.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012377.hg.1 |
NM_001076680.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012377.hg.1 |
NM_001076680.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012377.hg.1 |
NM_001076680.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012377.hg.1 |
NM_001076680.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012377.hg.1 |
NM_001076680.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700012377.hg.1 |
NM_001076680.1 |
Refseq |