Assay Details
| Gene Symbol: ZKSCAN8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278119.1 | NP_001265048.1 | 2 - 3 | 215 | 64 | |
| NM_001278121.1 | NP_001265050.1 | 2 - 3 | 148 | 64 | ||
| XM_011514873.2 | XP_011513175.1 | 2 - 3 | 307 | 64 | ||
| GenBank mRNA | AK302402.1 | - | 2 - 3 | 148 | 64 | |
| BC096282.3 | - | 2 - 3 | 187 | 40006900 | 64 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK302402.1, BC096282.3, NM_001278119.1, NM_001278121.1, XM_011514873.2 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000330236.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000330236.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000330236.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000330236.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000330236.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000330236.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000330236.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000330236.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000330236.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000330236.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000457389.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000457389.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000457389.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000457389.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000457389.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000457389.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000457389.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000457389.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000457389.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000536028.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000536028.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000536028.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000536028.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000536028.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000536028.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000536028.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000606198.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000606198.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000606198.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000606198.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000606198.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000606198.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000606198.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000606198.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000606198.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000606198.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600150096.hg.1 | ENST00000330236.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600150096.hg.1 | ENST00000457389.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600150096.hg.1 | ENST00000536028.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600150096.hg.1 | ENST00000606198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600150096.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600150096.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600150096.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600150096.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600150096.hg.1 | ENST00000330236.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600150096.hg.1 | ENST00000457389.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600150096.hg.1 | ENST00000536028.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600150096.hg.1 | ENST00000606198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600150096.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600150096.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600150096.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600150096.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600150096.hg.1 | TC0600007402.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600150094.hg.1 | ENST00000457389.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600150094.hg.1 | ENST00000536028.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600150094.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600150094.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600150094.hg.1 | ENST00000457389.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600150094.hg.1 | ENST00000536028.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600150094.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600150094.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600150094.hg.1 | TC0600007402.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000330236.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000330236.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000330236.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000330236.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000330236.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000330236.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000330236.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000330236.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000330236.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000330236.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000457389.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000457389.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000457389.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000457389.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000457389.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000457389.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000457389.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000457389.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000457389.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000536028.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000536028.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000536028.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000536028.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000536028.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000536028.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000536028.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000606198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000606198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000606198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000606198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000606198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000606198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000606198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000606198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000606198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000606198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600150093.hg.1 | ENST00000536028.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600150093.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600150093.hg.1 | ENST00000536028.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600150093.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600150093.hg.1 | TC0600007402.hg.1 | Clariom D TC |

ZKSCAN8
Genomic Map