Assay Details
| Gene Symbol: ZKSCAN8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278119.1 | NP_001265048.1 | 3 - 4 | 717 | 90 | |
| NM_006298.3 | NP_006289.2 | 2 - 3 | 623 | 90 | ||
| XM_011514870.2 | XP_011513172.1 | 2 - 3 | 712 | 90 | ||
| XM_017011266.1 | XP_016866755.1 | 2 - 3 | 627 | 90 | ||
| GenBank mRNA | AA453245.1 | - | 1 - 2 | 270 | 90 | |
| BC096282.3 | - | 3 - 4 | 689 | 40006900 | 90 | |
| BC096283.3 | - | 2 - 3 | 595 | 40006903 | 90 | |
| BC096284.3 | - | 2 - 3 | 595 | 40006905 | 90 | |
| BC113961.1 | - | 2 - 3 | 632 | 40022943 | 90 | |
| BC130032.1 | - | 2 - 3 | 613 | 40018337 | 90 | |
| BC152378.1 | - | 2 - 3 | 632 | 40018341 | 90 | |
| U57796.1 | - | 2 - 3 | 595 | 90 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06000276.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR06002294.hg.1 | ENST00000330236.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06002294.hg.1 | ENST00000457389.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06002294.hg.1 | ENST00000536028.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06002294.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06002294.hg.1 | TC06000276.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000330236.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000330236.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000330236.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000330236.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000330236.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000330236.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000330236.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000330236.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000330236.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000330236.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000457389.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000457389.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000457389.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000457389.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000457389.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000457389.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000457389.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000457389.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000457389.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000536028.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000536028.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000536028.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000536028.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000536028.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000536028.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000536028.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000606198.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000606198.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000606198.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000606198.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000606198.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000606198.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000606198.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000606198.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000606198.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | ENST00000606198.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007402.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600150099.hg.1 | ENST00000330236.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600150099.hg.1 | ENST00000457389.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600150099.hg.1 | ENST00000536028.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600150099.hg.1 | ENST00000606198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600150099.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600150099.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600150099.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600150099.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600150099.hg.1 | ENST00000330236.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600150099.hg.1 | ENST00000457389.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600150099.hg.1 | ENST00000536028.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600150099.hg.1 | ENST00000606198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600150099.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600150099.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600150099.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600150099.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600150099.hg.1 | TC0600007402.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000330236.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000330236.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000330236.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000330236.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000330236.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000330236.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000330236.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000330236.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000330236.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000330236.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000457389.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000457389.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000457389.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000457389.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000457389.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000457389.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000457389.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000457389.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000457389.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000536028.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000536028.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000536028.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000536028.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000536028.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000536028.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000536028.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000606198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000606198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000606198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000606198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000606198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000606198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000606198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000606198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000606198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | ENST00000606198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278119.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278121.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_001278122.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_006298.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007402.hg.1 | NM_006298.3 | Refseq |

ZKSCAN8
Genomic Map