| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
ENST00000330236.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
ENST00000330236.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
ENST00000330236.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
ENST00000330236.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
ENST00000330236.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
ENST00000330236.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
ENST00000330236.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
ENST00000330236.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
ENST00000330236.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
ENST00000330236.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
ENST00000457389.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
ENST00000457389.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
ENST00000457389.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
ENST00000457389.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
ENST00000457389.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
ENST00000457389.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
ENST00000457389.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
ENST00000457389.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
ENST00000457389.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
ENST00000536028.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
ENST00000536028.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
ENST00000536028.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
ENST00000536028.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
ENST00000536028.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
ENST00000536028.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
ENST00000536028.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
ENST00000606198.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
ENST00000606198.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
ENST00000606198.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
ENST00000606198.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
ENST00000606198.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
ENST00000606198.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
ENST00000606198.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
ENST00000606198.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
ENST00000606198.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
ENST00000606198.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
NM_001278119.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
NM_001278119.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
NM_001278119.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
NM_001278119.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
NM_001278119.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
NM_001278119.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
NM_001278119.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
NM_001278119.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
NM_001278119.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
NM_001278119.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
NM_001278119.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
NM_001278119.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
NM_001278121.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
NM_001278121.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
NM_001278121.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
NM_001278121.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
NM_001278121.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
NM_001278121.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
NM_001278121.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
NM_001278121.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
NM_001278121.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
NM_001278121.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
NM_001278121.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
NM_001278122.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
NM_001278122.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
NM_001278122.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
NM_001278122.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
NM_001278122.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
NM_001278122.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
NM_001278122.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
NM_001278122.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
NM_001278122.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
NM_001278122.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
NM_001278122.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
NM_006298.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
NM_006298.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
NM_006298.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
NM_006298.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
NM_006298.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
NM_006298.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
NM_006298.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
NM_006298.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
NM_006298.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
NM_006298.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007402.hg.1 |
NM_006298.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06002296.hg.1 |
ENST00000330236.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06002296.hg.1 |
ENST00000457389.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06002296.hg.1 |
ENST00000536028.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06002296.hg.1 |
NM_006298.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06002296.hg.1 |
TC06000276.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0600150101.hg.1 |
ENST00000330236.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600150101.hg.1 |
ENST00000457389.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600150101.hg.1 |
ENST00000536028.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600150101.hg.1 |
ENST00000606198.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600150101.hg.1 |
NM_001278119.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600150101.hg.1 |
NM_001278121.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600150101.hg.1 |
NM_001278122.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600150101.hg.1 |
NM_006298.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600150101.hg.1 |
ENST00000330236.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600150101.hg.1 |
ENST00000457389.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600150101.hg.1 |
ENST00000536028.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600150101.hg.1 |
ENST00000606198.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600150101.hg.1 |
NM_001278119.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600150101.hg.1 |
NM_001278121.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600150101.hg.1 |
NM_001278122.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600150101.hg.1 |
NM_006298.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600150101.hg.1 |
TC0600007402.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
ENST00000330236.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
ENST00000330236.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
ENST00000330236.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
ENST00000330236.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
ENST00000330236.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
ENST00000330236.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
ENST00000330236.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
ENST00000330236.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
ENST00000330236.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
ENST00000330236.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
ENST00000457389.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
ENST00000457389.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
ENST00000457389.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
ENST00000457389.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
ENST00000457389.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
ENST00000457389.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
ENST00000457389.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
ENST00000457389.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
ENST00000457389.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
ENST00000536028.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
ENST00000536028.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
ENST00000536028.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
ENST00000536028.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
ENST00000536028.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
ENST00000536028.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
ENST00000536028.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
ENST00000606198.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
ENST00000606198.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
ENST00000606198.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
ENST00000606198.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
ENST00000606198.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
ENST00000606198.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
ENST00000606198.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
ENST00000606198.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
ENST00000606198.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
ENST00000606198.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
NM_001278119.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
NM_001278119.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
NM_001278119.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
NM_001278119.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
NM_001278119.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
NM_001278119.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
NM_001278119.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
NM_001278119.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
NM_001278119.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
NM_001278119.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
NM_001278119.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
NM_001278119.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
NM_001278121.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
NM_001278121.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
NM_001278121.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
NM_001278121.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
NM_001278121.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
NM_001278121.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
NM_001278121.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
NM_001278121.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
NM_001278121.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
NM_001278121.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
NM_001278121.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
NM_001278122.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
NM_001278122.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
NM_001278122.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
NM_001278122.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
NM_001278122.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
NM_001278122.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
NM_001278122.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
NM_001278122.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
NM_001278122.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
NM_001278122.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
NM_001278122.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
NM_006298.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
NM_006298.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
NM_006298.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
NM_006298.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
NM_006298.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
NM_006298.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
NM_006298.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
NM_006298.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
NM_006298.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
NM_006298.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007402.hg.1 |
NM_006298.3 |
Refseq |